Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01441961_g1
          See other TXNDC15 GE Assays ›
          Gene Symbol:
          Txndc15
          Gene Name:
          thioredoxin domain containing 15
          Gene Aliases:
          RGD1304696
          Chromosome Location:
          Chr.17: 9429171 - 9441628 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01441961_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Txndc15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024998.1 NP_001020169.1 3 - 4 773 79
          XM_008771497.1 XP_008769719.1 3 - 4 881 79
          GenBank mRNA BC091340.1 - 3 - 4 773 7315216 79

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Txndc15

          Gene Ontology Categories:

          Function(s)

          cell redox homeostasis

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P457773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123423 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28793 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P457773 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P457773 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P457773 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P457773 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101778 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32078 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10797184 - -
          Affymetrix Rat Gene 1.0 ST Array 10797183 - -
          Affymetrix Rat Gene 1.0 ST Array 10797182 - -
          Affymetrix Rat Gene 1.0 ST Array 10797185 - -
          Affymetrix Rat Gene 1.0 ST Array 10797186 - -
          Affymetrix Rat Gene 1.1 ST Array 10797181 - -
          Rat Genome 230 2.0 Array 1367510_at - -
          Rat Genome 230 2.0 Array 1367510_PM_at - -
          Affymetrix HT Rat Focus Array 1367510_PM_at - -
          Affymetrix HT Rat Focus Array 1367510_at - -
          Affymetrix Rat Clariom S Assay PSR1700008940.rn.1 TC1700001301.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700008938.rn.1 TC1700001301.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700008939.rn.1 TC1700001301.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700008944.rn.1 TC1700001301.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700008943.rn.1 TC1700001301.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700008937.rn.1 TC1700001301.rn.1 RTA TC
          Rat Expression Set 230 1367510_PM_at - -
          Rat Expression Set 230 1367510_at - -
          Rat Expression Set 230 1367510_at - -
          Rat Expression Set 230 1367510_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10797181 - -
          Rat Genome U34 Set rc_AA849788_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21003924 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365031 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline