Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01446656_g1
          See other GSTA4 GE Assays ›
          Gene Symbol:
          Gsta4
          Gene Name:
          glutathione S-transferase alpha 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 85497557 - 85514732 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01446656_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gsta4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106840.1 NP_001100310.1 205 109
          GenBank mRNA BC157819.1 - 175 5621777 109
          FJ179399.1 - 2 - 3 138 109
          FQ230022.1 - 185 109
          FQ233696.1 - 187 109
          FQ233840.1 - 187 109
          X62660.1 - 166 109

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          glutathione metabolic process
          xenobiotic metabolic process
          response to herbicide
          response to zinc ion
          response to nicotine
          cellular response to lithium ion
          glutathione transferase activity
          drug binding
          protein homodimerization activity
          glutathione binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P019106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461049 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147040 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91397 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019106 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019106 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019106 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P531078 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24292 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10884920 - -
          Affymetrix Rat Gene 1.0 ST Array 10884925 - -
          Affymetrix Rat Gene 1.0 ST Array 10911799 - -
          Affymetrix Rat Gene 1.0 ST Array 10911798 - -
          Affymetrix Rat Gene 1.0 ST Array 10911800 - -
          Affymetrix Rat Gene 1.0 ST Array 10884919 - -
          Affymetrix Rat Gene 1.0 ST Array 10911801 - -
          Affymetrix Rat Gene 1.1 ST Array 10884924 - -
          Affymetrix Rat Gene 1.1 ST Array 10911797 - -
          Affymetrix Rat Gene 1.1 ST Array 10884917 - -
          Rat Genome 230 2.0 Array 1372297_at - -
          Rat Genome 230 2.0 Array 1372297_PM_at - -
          Affymetrix HT Rat Focus Array 1372297_at - -
          Affymetrix HT Rat Focus Array 1372297_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800008484.rn.1 TC0800001158.rn.1 RTA TC
          Rat Expression Set 230 1372297_PM_at - -
          Rat Expression Set 230 1372297_at - -
          Affymetrix Rat Gene 1.0 ST Array 10911797 - -
          Affymetrix Rat Gene 1.0 ST Array 10884917 - -
          Rat Genome U34 Set X62660mRNA_g_at - -
          Rat Genome U34 Set X62660mRNA_at - -
          Rat Genome U34 Set rc_AI234527_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21555860 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352039 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-xpdfw:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline