Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01449516_m1
          See other TANC1 GE Assays ›
          Gene Symbol:
          Tanc1
          Gene Name:
          tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1
          Gene Aliases:
          Tanc
          Chromosome Location:
          Chr.3: 45718581 - 45917687 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01449516_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tanc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001002854.1 NP_001002854.1 25 - 26 4243 95
          XM_006234204.1 XP_006234266.1 24 - 25 4162 95
          XM_006234205.2 XP_006234267.1 23 - 24 6398 95
          XM_006234207.2 XP_006234269.1 20 - 21 4360 95
          XM_008761838.1 XP_008760060.1 23 - 24 4576 95
          XM_008761839.1 XP_008760061.1 23 - 24 4862 95
          XM_008761840.1 XP_008760062.1 21 - 22 5494 95
          GenBank mRNA AB098072.1 - 25 - 26 4243 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000649.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tanc1

          Gene Ontology Categories:

          Function(s) Process(es)

          myoblast fusion
          visual learning
          dendritic spine maintenance
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P148473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 693974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600784 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126109 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18848 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106665 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44183 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6808 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64802 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73660 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128720 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26397 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113314 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160734 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28177 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141309 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48179 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26076 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139121 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113433 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86593 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89528 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42747 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148473 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P148473 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P148473 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P182336 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42982 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92381 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2764 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90726 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102353 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45645 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78725 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19388 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43316 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30847 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80936 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30897 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81162 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10836369 - -
          Affymetrix Rat Gene 1.0 ST Array 10836380 - -
          Affymetrix Rat Gene 1.0 ST Array 10836372 - -
          Affymetrix Rat Gene 1.0 ST Array 10836366 - -
          Affymetrix Rat Gene 1.0 ST Array 10836356 - -
          Affymetrix Rat Gene 1.0 ST Array 10836364 - -
          Affymetrix Rat Gene 1.0 ST Array 10836359 - -
          Affymetrix Rat Gene 1.0 ST Array 10836363 - -
          Affymetrix Rat Gene 1.0 ST Array 10836375 - -
          Affymetrix Rat Gene 1.0 ST Array 10836379 - -
          Affymetrix Rat Gene 1.0 ST Array 10836374 - -
          Affymetrix Rat Gene 1.0 ST Array 10836370 - -
          Affymetrix Rat Gene 1.0 ST Array 10836368 - -
          Affymetrix Rat Gene 1.0 ST Array 10836371 - -
          Affymetrix Rat Gene 1.0 ST Array 10836362 - -
          Affymetrix Rat Gene 1.0 ST Array 10836358 - -
          Affymetrix Rat Gene 1.0 ST Array 10836367 - -
          Affymetrix Rat Gene 1.0 ST Array 10836373 - -
          Affymetrix Rat Gene 1.0 ST Array 10836365 - -
          Affymetrix Rat Gene 1.0 ST Array 10836381 - -
          Affymetrix Rat Gene 1.0 ST Array 10836360 - -
          Affymetrix Rat Gene 1.0 ST Array 10836376 - -
          Affymetrix Rat Gene 1.0 ST Array 10836357 - -
          Affymetrix Rat Gene 1.0 ST Array 10836355 - -
          Affymetrix Rat Gene 1.0 ST Array 10836382 - -
          Affymetrix Rat Gene 1.1 ST Array 10836354 - -
          Rat Genome 230 2.0 Array 1373922_PM_at - -
          Rat Genome 230 2.0 Array 1373922_at - -
          Affymetrix Rat Clariom S Assay PSR0300004976.rn.1 TC0300000649.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10836354 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21585471 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356102 - -
          RatRef-12 Expression BeadChip ILMN_56181 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline