Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01450134_g1
          See other HNRPD GE Assays ›
          Gene Symbol:
          Hnrpd
          Gene Name:
          heterogeneous nuclear ribonucleoprotein D
          Gene Aliases:
          Auf1, Hnrnpd
          Chromosome Location:
          Chr.14: 11256163 - 11274684 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01450134_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hnrpd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001082539.1 NP_001076008.1 2 - 3 680 159
          NM_001082540.1 NP_001076009.1 3 - 4 737 159
          NM_001082541.1 NP_001076010.1 2 - 3 680 159
          NM_024404.2 NP_077380.2 3 - 4 737 159
          GenBank mRNA AB046615.1 - 3 - 4 501 159
          AB046616.1 - 2 - 3 395 159
          AB046617.1 - 3 - 4 452 159
          AB046618.1 - 2 - 3 395 159

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hnrpd

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, exonucleolytic
          nucleocytoplasmic transport
          regulation of circadian rhythm
          regulation of mRNA stability
          positive regulation of translation
          mRNA stabilization
          circadian regulation of translation
          nucleotide binding
          nucleic acid binding
          RNA binding
          mRNA binding
          telomeric DNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P363156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610199 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173696 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26452 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P363156 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P363156 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P363156 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1022421 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P363156 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701734980 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79190 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2009 - -
          Agilent Rat cDNA Microarray (G4105A) 701734980 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10771478 - -
          Affymetrix Rat Gene 1.0 ST Array 10771476 - -
          Affymetrix Rat Gene 1.0 ST Array 10771481 - -
          Affymetrix Rat Gene 1.0 ST Array 10771484 - -
          Affymetrix Rat Gene 1.0 ST Array 10771477 - -
          Affymetrix Rat Gene 1.0 ST Array 10771482 - -
          Affymetrix Rat Gene 1.0 ST Array 10771479 - -
          Affymetrix Rat Gene 1.0 ST Array 10771480 - -
          Affymetrix Rat Gene 1.0 ST Array 10771483 - -
          Affymetrix Rat Gene 1.1 ST Array 10771475 - -
          Rat Genome 230 2.0 Array 1369971_a_at - -
          Rat Genome 230 2.0 Array 1369971_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369971_a_at - -
          Affymetrix HT Rat Focus Array 1369971_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR1400001519.rn.1 TC1400000188.rn.1 RTA TC
          Rat Expression Set 230 1369971_PM_a_at - -
          Rat Expression Set 230 1369971_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10771475 - -
          Rat Genome U34 Set rc_AA945952_g_at - -
          Rat Genome U34 Set rc_AA957413_at - -
          Rat Genome U34 Set rc_AA945952_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20915510 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356233 - -
          RatRef-12 Expression BeadChip ILMN_48437 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline