Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01450146_m1
          See other PAQR3 GE Assays ›
          Gene Symbol:
          Paqr3
          Gene Name:
          progestin and adipoQ receptor family member III
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 14107518 - 14126464 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01450146_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Paqr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012033.1 NP_001012033.1 5 - 6 914 104
          XM_008770025.1 XP_008768247.1 5 - 6 976 104
          GenBank mRNA BC079418.1 - 5 - 6 914 7113653 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000210.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Paqr3

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein phosphorylation
          negative regulation of neuron projection development
          negative regulation of peptidyl-serine phosphorylation
          protein localization to Golgi apparatus
          negative regulation of MAP kinase activity
          receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P036365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556028 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148520 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21490 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109529 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036365 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P036365 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P036365 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P499056 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31768 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12235 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83777 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10771553 - -
          Affymetrix Rat Gene 1.0 ST Array 10771551 - -
          Affymetrix Rat Gene 1.0 ST Array 10771550 - -
          Affymetrix Rat Gene 1.0 ST Array 10771555 - -
          Affymetrix Rat Gene 1.0 ST Array 10771554 - -
          Affymetrix Rat Gene 1.0 ST Array 10771552 - -
          Affymetrix Rat Gene 1.0 ST Array 10771549 - -
          Affymetrix Rat Gene 1.1 ST Array 10771548 - -
          Affymetrix Rat Clariom S Assay PSR1400001719.rn.1 TC1400000210.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001721.rn.1 TC1400000210.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400001722.rn.1 TC1400000210.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10771548 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21105485 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2040870 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline