Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01451522_m1
          See other ATG2A GE Assays ›
          Gene Symbol:
          Atg2a
          Gene Name:
          autophagy related 2A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 221612584 - 221634799 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01451522_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atg2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109545.1 NP_001103015.1 1 - 2 302 64
          XM_008760180.1 XP_008758402.1 1 - 2 283 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003098.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s)

          autophagosome assembly
          mitophagy
          cellular response to nitrogen starvation
          nucleophagy

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P19531 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P152622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374689 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137725 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158716 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152622 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19531 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152622 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19531 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P152622 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P19531 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P427648 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70502 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100202 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713227 - -
          Affymetrix Rat Gene 1.0 ST Array 10713231 - -
          Affymetrix Rat Gene 1.0 ST Array 10713262 - -
          Affymetrix Rat Gene 1.0 ST Array 10713263 - -
          Affymetrix Rat Gene 1.0 ST Array 10713250 - -
          Affymetrix Rat Gene 1.0 ST Array 10713228 - -
          Affymetrix Rat Gene 1.0 ST Array 10713230 - -
          Affymetrix Rat Gene 1.0 ST Array 10713257 - -
          Affymetrix Rat Gene 1.0 ST Array 10713252 - -
          Affymetrix Rat Gene 1.0 ST Array 10713258 - -
          Affymetrix Rat Gene 1.0 ST Array 10713255 - -
          Affymetrix Rat Gene 1.0 ST Array 10713240 - -
          Affymetrix Rat Gene 1.0 ST Array 10713235 - -
          Affymetrix Rat Gene 1.0 ST Array 10713233 - -
          Affymetrix Rat Gene 1.0 ST Array 10713224 - -
          Affymetrix Rat Gene 1.0 ST Array 10713264 - -
          Affymetrix Rat Gene 1.0 ST Array 10713254 - -
          Affymetrix Rat Gene 1.0 ST Array 10713261 - -
          Affymetrix Rat Gene 1.0 ST Array 10713265 - -
          Affymetrix Rat Gene 1.0 ST Array 10713232 - -
          Affymetrix Rat Gene 1.0 ST Array 10713253 - -
          Affymetrix Rat Gene 1.0 ST Array 10713225 - -
          Affymetrix Rat Gene 1.0 ST Array 10713260 - -
          Affymetrix Rat Gene 1.0 ST Array 10713223 - -
          Affymetrix Rat Gene 1.0 ST Array 10713237 - -
          Affymetrix Rat Gene 1.0 ST Array 10713259 - -
          Affymetrix Rat Gene 1.0 ST Array 10713246 - -
          Affymetrix Rat Gene 1.0 ST Array 10713247 - -
          Affymetrix Rat Gene 1.0 ST Array 10713266 - -
          Affymetrix Rat Gene 1.0 ST Array 10713238 - -
          Affymetrix Rat Gene 1.0 ST Array 10713226 - -
          Affymetrix Rat Gene 1.0 ST Array 10713239 - -
          Affymetrix Rat Gene 1.0 ST Array 10713234 - -
          Affymetrix Rat Gene 1.0 ST Array 10713243 - -
          Affymetrix Rat Gene 1.0 ST Array 10713251 - -
          Affymetrix Rat Gene 1.0 ST Array 10713245 - -
          Affymetrix Rat Gene 1.0 ST Array 10713236 - -
          Affymetrix Rat Gene 1.0 ST Array 10713244 - -
          Affymetrix Rat Gene 1.0 ST Array 10713249 - -
          Affymetrix Rat Gene 1.0 ST Array 10713248 - -
          Affymetrix Rat Gene 1.0 ST Array 10713242 - -
          Affymetrix Rat Gene 1.0 ST Array 10713241 - -
          Affymetrix Rat Gene 1.0 ST Array 10713229 - -
          Affymetrix Rat Gene 1.1 ST Array 10713222 - -
          Rat Genome 230 2.0 Array 1377665_at - -
          Rat Genome 230 2.0 Array 1377665_PM_at - -
          Affymetrix HT Rat Focus Array 1377665_at - -
          Affymetrix HT Rat Focus Array 1377665_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100025156.rn.1 TC0100003098.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025155.rn.1 TC0100003098.rn.1 RTA TC
          Rat Expression Set 230 1377665_PM_at - -
          Rat Expression Set 230 1377665_at - -
          Affymetrix Rat Gene 1.0 ST Array 10713222 - -
          Rat Genome U34 Set rc_AA998979_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22007796 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline