Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01451817_m1
          See other ZFP398 GE Assays ›
          Gene Symbol:
          Zfp398
          Gene Name:
          zinc finger protein 398
          Gene Aliases:
          RGD1562129
          Chromosome Location:
          Chr.4: 77519023 - 77546517 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01451817_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp398
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109230.1 NP_001102700.1 5 - 6 1004 83
          XM_008762904.1 XP_008761126.1 4 - 5 613 83
          XM_008762905.1 XP_008761127.1 4 - 5 495 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000832.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P262587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639562 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136581 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P161483 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P161483 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P161483 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57442 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10855193 - -
          Affymetrix Rat Gene 1.0 ST Array 10855190 - -
          Affymetrix Rat Gene 1.0 ST Array 10855188 - -
          Affymetrix Rat Gene 1.0 ST Array 10855192 - -
          Affymetrix Rat Gene 1.0 ST Array 10855189 - -
          Affymetrix Rat Gene 1.0 ST Array 10855191 - -
          Affymetrix Rat Gene 1.1 ST Array 10855187 - -
          Rat Genome 230 2.0 Array 1397447_at - -
          Rat Genome 230 2.0 Array 1397447_PM_at - -
          Affymetrix HT Rat Focus Array 1397447_at - -
          Affymetrix HT Rat Focus Array 1397447_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400005821.rn.1 TC0400000832.rn.1 RTA TC
          Rat Expression Set 230 1397447_at - -
          Rat Expression Set 230 1397447_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10855187 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22307078 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650497 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline