Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01451853_m1
          See other RARRES2 GE Assays ›
          Gene Symbol:
          Rarres2
          Gene Name:
          retinoic acid receptor responder (tazarotene induced) 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 78205809 - 78208940 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01451853_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rarres2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013427.1 NP_001013445.1 3 - 4 369 116
          XM_006236415.2 XP_006236477.1 3 - 4 396 116
          XM_006236416.1 XP_006236478.1 3 - 4 457 116
          GenBank mRNA BC091177.1 - 3 - 4 369 7323591 116
          FQ209660.1 - 3 - 4 384 116
          FQ209771.1 - 3 - 4 383 116
          FQ210577.1 - 3 - 4 384 116
          FQ228347.1 - 3 - 4 418 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002841.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rarres2

          Gene Ontology Categories:

          Function(s) Process(es)

          retinoid metabolic process
          in utero embryonic development
          positive regulation of protein phosphorylation
          inflammatory response
          positive regulation of macrophage chemotaxis
          positive regulation of fat cell differentiation
          embryonic digestive tract development
          brown fat cell differentiation
          positive regulation of chemotaxis
          regulation of lipid catabolic process
          positive regulation of glucose import in response to insulin stimulus
          receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P281481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784066 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P281481 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P281481 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P281481 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P281481 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10862409 - -
          Affymetrix Rat Gene 1.0 ST Array 10862407 - -
          Affymetrix Rat Gene 1.0 ST Array 10862408 - -
          Affymetrix Rat Gene 1.0 ST Array 10862406 - -
          Affymetrix Rat Gene 1.0 ST Array 10862404 - -
          Affymetrix Rat Gene 1.0 ST Array 10862405 - -
          Affymetrix Rat Gene 1.1 ST Array 10862403 - -
          Rat Genome 230 2.0 Array 1393618_at - -
          Rat Genome 230 2.0 Array 1393618_PM_at - -
          Rat Genome 230 2.0 Array 1371691_PM_at - -
          Rat Genome 230 2.0 Array 1371691_at - -
          Affymetrix HT Rat Focus Array 1371691_at - -
          Affymetrix HT Rat Focus Array 1371691_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400020237.rn.1 TC0400002841.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020243.rn.1 TC0400002841.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020239.rn.1 TC0400002841.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020242.rn.1 TC0400002841.rn.1 RTA TC
          Rat Expression Set 230 1371691_at - -
          Rat Expression Set 230 1371691_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10862403 - -
          Rat Genome U34 Set rc_AA851273_g_at - -
          Rat Genome U34 Set rc_AA851273_at - -
          Rat Genome U34 Set rc_AI176061_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21747535 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370339 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline