Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01452182_m1
          See other CARF GE Assays ›
          Gene Symbol:
          Carf
          Gene Name:
          calcium responsive transcription factor
          Gene Aliases:
          Als2cr8
          Chromosome Location:
          Chr.9: 66878551 - 66928658 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01452182_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Carf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106915.1 NP_001100385.1 12 - 13 1372 85
          XM_006244969.1 XP_006245031.1 9 - 10 1234 85
          XM_006244970.2 XP_006245032.1 10 - 11 1520 85
          XM_006244971.1 XP_006245033.1 8 - 9 1075 85
          XM_006244973.2 XP_006245035.1 7 - 8 796 85
          XM_006244974.2 XP_006245036.1 8 - 9 1760 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000742.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          cellular response to potassium ion
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of transcription from RNA polymerase II promoter in response to calcium ion
          cellular response to calcium ion
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P23451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843506 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10719 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180508 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146328 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36492 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21650 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P23451 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P23451 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P23451 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P23451 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85899 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55010 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74040 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10923721 - -
          Affymetrix Rat Gene 1.0 ST Array 10923714 - -
          Affymetrix Rat Gene 1.0 ST Array 10923711 - -
          Affymetrix Rat Gene 1.0 ST Array 10923712 - -
          Affymetrix Rat Gene 1.0 ST Array 10923720 - -
          Affymetrix Rat Gene 1.0 ST Array 10923715 - -
          Affymetrix Rat Gene 1.0 ST Array 10923723 - -
          Affymetrix Rat Gene 1.0 ST Array 10923722 - -
          Affymetrix Rat Gene 1.0 ST Array 10923718 - -
          Affymetrix Rat Gene 1.0 ST Array 10923724 - -
          Affymetrix Rat Gene 1.0 ST Array 10923717 - -
          Affymetrix Rat Gene 1.0 ST Array 10923719 - -
          Affymetrix Rat Gene 1.0 ST Array 10923726 - -
          Affymetrix Rat Gene 1.0 ST Array 10923725 - -
          Affymetrix Rat Gene 1.0 ST Array 10923716 - -
          Affymetrix Rat Gene 1.1 ST Array 10923710 - -
          Affymetrix Rat Clariom S Assay PSR0900006056.rn.1 TC0900000742.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900006057.rn.1 TC0900000742.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10923710 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22424269 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371781 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline