Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01453631_m1
          See other TECPR1 GE Assays ›
          Gene Symbol:
          Tecpr1
          Gene Name:
          tectonin beta-propeller repeat containing 1
          Gene Aliases:
          RGD1306873
          Chromosome Location:
          Chr.12: 12374790 - 12402547 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01453631_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tecpr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037191.1 NP_001032268.1 18 - 19 2759 87
          GenBank mRNA BC103478.1 - 18 - 19 2759 7937275 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000209.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tecpr1

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagy
          autophagosome maturation
          phosphatidylinositol-3-phosphate binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P000824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669682 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118706 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66224 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93079 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000824 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P000824 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P000824 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21138 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P233988 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8742 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32281 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69333 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10756649 - -
          Affymetrix Rat Gene 1.0 ST Array 10756640 - -
          Affymetrix Rat Gene 1.0 ST Array 10756654 - -
          Affymetrix Rat Gene 1.0 ST Array 10756660 - -
          Affymetrix Rat Gene 1.0 ST Array 10756645 - -
          Affymetrix Rat Gene 1.0 ST Array 10756639 - -
          Affymetrix Rat Gene 1.0 ST Array 10756643 - -
          Affymetrix Rat Gene 1.0 ST Array 10756647 - -
          Affymetrix Rat Gene 1.0 ST Array 10756651 - -
          Affymetrix Rat Gene 1.0 ST Array 10756642 - -
          Affymetrix Rat Gene 1.0 ST Array 10756657 - -
          Affymetrix Rat Gene 1.0 ST Array 10756641 - -
          Affymetrix Rat Gene 1.0 ST Array 10756644 - -
          Affymetrix Rat Gene 1.0 ST Array 10756656 - -
          Affymetrix Rat Gene 1.0 ST Array 10756646 - -
          Affymetrix Rat Gene 1.0 ST Array 10756661 - -
          Affymetrix Rat Gene 1.0 ST Array 10756662 - -
          Affymetrix Rat Gene 1.0 ST Array 10756652 - -
          Affymetrix Rat Gene 1.0 ST Array 10756653 - -
          Affymetrix Rat Gene 1.0 ST Array 10756659 - -
          Affymetrix Rat Gene 1.0 ST Array 10756650 - -
          Affymetrix Rat Gene 1.0 ST Array 10756648 - -
          Affymetrix Rat Gene 1.0 ST Array 10756658 - -
          Affymetrix Rat Gene 1.0 ST Array 10756655 - -
          Affymetrix Rat Gene 1.1 ST Array 10756638 - -
          Rat Genome 230 2.0 Array 1373112_at - -
          Rat Genome 230 2.0 Array 1375656_PM_at - -
          Rat Genome 230 2.0 Array 1375656_at - -
          Rat Genome 230 2.0 Array 1373112_PM_at - -
          Affymetrix HT Rat Focus Array 1375656_PM_at - -
          Affymetrix HT Rat Focus Array 1375656_at - -
          Affymetrix HT Rat Focus Array 1373112_at - -
          Affymetrix HT Rat Focus Array 1373112_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200001457.rn.1 TC1200000209.rn.1 RTA TC
          Rat Expression Set 230 1375656_PM_at - -
          Rat Expression Set 230 1375656_at - -
          Rat Expression Set 230 1373112_PM_at - -
          Rat Expression Set 230 1373112_at - -
          Affymetrix Rat Gene 1.0 ST Array 10756638 - -
          Rat Genome U34 Set rc_AI043714_at - -
          Rat Genome U34 Set rc_AI112620_at - -
          Rat Genome U34 Set rc_AI230528_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650265 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline