Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01456688_m1
          See other NLGN2 GE Assays ›
          Gene Symbol:
          Nlgn2
          Gene Name:
          neuroligin 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 56431586 - 56444847 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01456688_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nlgn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053992.1 NP_446444.1 7 - 8 2180 59
          GenBank mRNA U41662.1 - 7 - 8 2180 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003103.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nlgn2

          Panther Classification:

          Molecular Function -

          cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          thigmotaxis
          regulation of respiratory gaseous exchange by neurological system process
          cell adhesion
          neuron cell-cell adhesion
          synapse assembly
          jump response
          metabolic process
          sensory perception of pain
          positive regulation of synaptic transmission, GABAergic
          social behavior
          protein localization to synapse
          locomotory exploration behavior
          modulation of synaptic transmission
          synapse organization
          neuromuscular process controlling balance
          positive regulation of synapse assembly
          positive regulation of synaptic transmission, glutamatergic
          terminal button organization
          postsynaptic membrane assembly
          presynaptic membrane assembly
          gephyrin clustering involved in postsynaptic density assembly
          postsynaptic density protein 95 clustering
          positive regulation of inhibitory postsynaptic potential
          regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
          positive regulation of excitatory postsynaptic potential
          receptor activity
          neurexin family protein binding
          cell adhesion molecule binding
          carboxylic ester hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P030188 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 942332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93710 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77488 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13011 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030188 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030188 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13011 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P030188 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13011 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13011 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P299542 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44197 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70342 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10744247 - -
          Affymetrix Rat Gene 1.0 ST Array 10744252 - -
          Affymetrix Rat Gene 1.0 ST Array 10744249 - -
          Affymetrix Rat Gene 1.0 ST Array 10744251 - -
          Affymetrix Rat Gene 1.0 ST Array 10744253 - -
          Affymetrix Rat Gene 1.0 ST Array 10744246 - -
          Affymetrix Rat Gene 1.0 ST Array 10744250 - -
          Affymetrix Rat Gene 1.0 ST Array 10744248 - -
          Affymetrix Rat Gene 1.1 ST Array 10744245 - -
          Rat Genome 230 2.0 Array 1397705_PM_at - -
          Rat Genome 230 2.0 Array 1368895_PM_at - -
          Rat Genome 230 2.0 Array 1397705_at - -
          Rat Genome 230 2.0 Array 1368895_at - -
          Affymetrix HT Rat Focus Array 1368895_at - -
          Affymetrix HT Rat Focus Array 1368895_PM_at - -
          Affymetrix HT Rat Focus Array 1397705_at - -
          Affymetrix HT Rat Focus Array 1397705_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000027679.rn.1 TC1000003103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000027680.rn.1 TC1000003103.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000027681.rn.1 TC1000003103.rn.1 RTA TC
          Rat Expression Set 230 1368895_PM_at - -
          Rat Expression Set 230 1368895_at - -
          Rat Expression Set 230 1397705_at - -
          Rat Expression Set 230 1397705_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10744245 - -
          Rat Genome U34 Set U41662_at - -
          Rat Genome U34 Set U41662_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21503386 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365568 - -
          RatRef-12 Expression BeadChip ILMN_63980 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline