Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01458195_m1
          See other BRINP1 GE Assays ›
          Gene Symbol:
          Brinp1
          Gene Name:
          bone morphogenetic protein/retinoic acid inducible neural-specific 1
          Gene Aliases:
          Dbc1, Dbccr1
          Chromosome Location:
          Chr.5: 84919418 - 85123879 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01458195_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Brinp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080482.2 NP_536730.1 5 - 6 980 71
          XM_006238272.2 XP_006238334.1 6 - 7 1398 71
          GenBank mRNA AB051356.1 - 5 - 6 981 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500002931.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Brinp1

          Gene Ontology Categories:

          Function(s)

          cell cycle arrest
          cell death
          positive regulation of neuron differentiation
          negative regulation of cell cycle
          negative regulation of mitotic cell cycle
          cellular response to retinoic acid

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P161220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32309 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89935 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144263 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51604 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154540 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27835 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99680 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81333 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120685 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89884 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166998 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99580 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161421 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75436 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99205 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131202 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27687 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P161220 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P161220 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P161220 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P161220 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99282 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37666 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59830 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63363 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48300 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55252 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57249 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29632 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5802 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86688 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10877579 - -
          Affymetrix Rat Gene 1.0 ST Array 10877576 - -
          Affymetrix Rat Gene 1.0 ST Array 10877575 - -
          Affymetrix Rat Gene 1.0 ST Array 10877574 - -
          Affymetrix Rat Gene 1.0 ST Array 10877577 - -
          Affymetrix Rat Gene 1.0 ST Array 10877580 - -
          Affymetrix Rat Gene 1.0 ST Array 10877578 - -
          Affymetrix Rat Gene 1.1 ST Array 10877573 - -
          Rat Genome 230 2.0 Array 1369408_at - -
          Rat Genome 230 2.0 Array 1369408_PM_at - -
          Affymetrix HT Rat Focus Array 1369408_at - -
          Affymetrix HT Rat Focus Array 1369408_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500021841.rn.1 TC0500002931.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500021842.rn.1 TC0500002931.rn.1 RTA TC
          Rat Expression Set 230 1369408_PM_at - -
          Rat Expression Set 230 1369408_at - -
          Affymetrix Rat Gene 1.0 ST Array 10877573 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365134 - -
          RatRef-12 Expression BeadChip ILMN_58992 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline