Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01460970_m1
          See other CDC27 GE Assays ›
          Gene Symbol:
          Cdc27
          Gene Name:
          cell division cycle 27
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 92551754 - 92602217 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01460970_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdc27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024793.1 NP_001019964.1 18 - 19 2470 79
          XM_006247533.2 XP_006247595.1 17 - 18 4191 79
          XM_006247534.2 XP_006247596.1 17 - 18 4188 79
          XM_006247535.2 XP_006247597.1 18 - 19 2607 79
          XM_006247536.2 XP_006247598.1 16 - 17 4039 79
          XM_006247537.1 XP_006247599.1 15 - 16 2139 79
          XM_008768295.1 XP_008766517.1 17 - 18 2460 79
          GenBank mRNA BC097475.1 - 18 - 19 2470 7457819 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003921.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdc27

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          metaphase/anaphase transition of mitotic cell cycle
          protein K11-linked ubiquitination
          protein phosphatase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P183220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623150 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903575 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156814 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99618 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119071 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41878 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64988 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P183220 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P183220 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P183220 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P183220 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1032285 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7997 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80382 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42644 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32060 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10748156 - -
          Affymetrix Rat Gene 1.0 ST Array 10748158 - -
          Affymetrix Rat Gene 1.0 ST Array 10748169 - -
          Affymetrix Rat Gene 1.0 ST Array 10748160 - -
          Affymetrix Rat Gene 1.0 ST Array 10748173 - -
          Affymetrix Rat Gene 1.0 ST Array 10748165 - -
          Affymetrix Rat Gene 1.0 ST Array 10748167 - -
          Affymetrix Rat Gene 1.0 ST Array 10748172 - -
          Affymetrix Rat Gene 1.0 ST Array 10748164 - -
          Affymetrix Rat Gene 1.0 ST Array 10748166 - -
          Affymetrix Rat Gene 1.0 ST Array 10748159 - -
          Affymetrix Rat Gene 1.0 ST Array 10748162 - -
          Affymetrix Rat Gene 1.0 ST Array 10748168 - -
          Affymetrix Rat Gene 1.0 ST Array 10748161 - -
          Affymetrix Rat Gene 1.0 ST Array 10748170 - -
          Affymetrix Rat Gene 1.0 ST Array 10748163 - -
          Affymetrix Rat Gene 1.0 ST Array 10748157 - -
          Affymetrix Rat Gene 1.0 ST Array 10748155 - -
          Affymetrix Rat Gene 1.0 ST Array 10748171 - -
          Affymetrix Rat Gene 1.1 ST Array 10748154 - -
          Rat Genome 230 2.0 Array 1382664_at - -
          Rat Genome 230 2.0 Array 1382664_PM_at - -
          Affymetrix HT Rat Focus Array 1382664_PM_at - -
          Affymetrix HT Rat Focus Array 1382664_at - -
          Affymetrix Rat Clariom S Assay PSR1000035599.rn.1 TC1000003921.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000035598.rn.1 TC1000003921.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000035596.rn.1 TC1000003921.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000035601.rn.1 TC1000003921.rn.1 RTA TC
          Rat Expression Set 230 1382664_at - -
          Rat Expression Set 230 1382664_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10748154 - -
          Rat Genome U34 Set rc_AI045565_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22385499 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359941 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline