Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01462252_m1
          See other MCCC1 GE Assays ›
          Gene Symbol:
          Mccc1
          Gene Name:
          methylcrotonoyl-CoA carboxylase 1 (alpha)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 122550777 - 122690540 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01462252_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mccc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009653.1 NP_001009653.1 18 - 19 2058 131
          GenBank mRNA BC088473.1 - 18 - 19 2058 7128872 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003505.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mccc1

          Panther Classification:

          Molecular Function -

          ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          leucine catabolic process
          metabolic process
          biotin carboxylase activity
          methylcrotonoyl-CoA carboxylase activity
          ATP binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 152017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155570 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133395 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91177 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150691 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43059 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14764 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44113 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127701 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14611 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14611 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14611 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14611 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59188 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78408 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67686 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10822817 - -
          Affymetrix Rat Gene 1.0 ST Array 10822804 - -
          Affymetrix Rat Gene 1.0 ST Array 10822815 - -
          Affymetrix Rat Gene 1.0 ST Array 10822811 - -
          Affymetrix Rat Gene 1.0 ST Array 10822800 - -
          Affymetrix Rat Gene 1.0 ST Array 10822805 - -
          Affymetrix Rat Gene 1.0 ST Array 10822803 - -
          Affymetrix Rat Gene 1.0 ST Array 10822808 - -
          Affymetrix Rat Gene 1.0 ST Array 10822806 - -
          Affymetrix Rat Gene 1.0 ST Array 10822810 - -
          Affymetrix Rat Gene 1.0 ST Array 10822814 - -
          Affymetrix Rat Gene 1.0 ST Array 10822818 - -
          Affymetrix Rat Gene 1.0 ST Array 10822813 - -
          Affymetrix Rat Gene 1.0 ST Array 10822819 - -
          Affymetrix Rat Gene 1.0 ST Array 10822801 - -
          Affymetrix Rat Gene 1.0 ST Array 10822809 - -
          Affymetrix Rat Gene 1.0 ST Array 10822802 - -
          Affymetrix Rat Gene 1.0 ST Array 10822812 - -
          Affymetrix Rat Gene 1.0 ST Array 10822807 - -
          Affymetrix Rat Gene 1.0 ST Array 10822816 - -
          Affymetrix Rat Gene 1.1 ST Array 10822799 - -
          Rat Genome 230 2.0 Array 1376852_PM_at - -
          Rat Genome 230 2.0 Array 1395844_PM_at - -
          Rat Genome 230 2.0 Array 1395844_at - -
          Rat Genome 230 2.0 Array 1376852_at - -
          Affymetrix HT Rat Focus Array 1376852_PM_at - -
          Affymetrix HT Rat Focus Array 1376852_at - -
          Affymetrix Rat Clariom S Assay PSR0200023805.rn.1 TC0200003505.rn.1 RTA TC
          Rat Expression Set 230 1376852_at - -
          Rat Expression Set 230 1376852_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10822799 - -
          Rat Genome U34 Set rc_AI105272_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21254188 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530393 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365662 - -
          RatRef-12 Expression BeadChip ILMN_160890 - -
          RatRef-12 Expression BeadChip ILMN_62535 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline