Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01465024_m1
          See other PKD1 GE Assays ›
          Gene Symbol:
          Pkd1
          Gene Name:
          polycystic kidney disease 1 homolog (human)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 13914492 - 13962008 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01465024_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pkd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001257352.1 NP_001244281.1 16 - 17 7051 54
          XM_008767538.1 XP_008765760.1 16 - 17 7773 54
          XM_008767539.1 XP_008765761.1 16 - 17 7773 54
          XM_008767540.1 XP_008765762.1 16 - 17 7773 54
          XM_008767541.1 XP_008765763.1 16 - 17 7772 54
          GenBank mRNA AF277452.1 - 16 - 17 7024 54

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000213.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          cartilage condensation
          blood vessel development
          in utero embryonic development
          kidney development
          liver development
          embryonic placenta development
          protein export from nucleus
          nitrogen compound metabolic process
          calcium ion transport
          cell cycle arrest
          positive regulation of cytosolic calcium ion concentration
          JAK-STAT cascade
          heart development
          single organismal cell-cell adhesion
          peptidyl-serine phosphorylation
          spinal cord development
          neural tube development
          establishment of cell polarity
          regulation of cell adhesion
          positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
          positive regulation of protein binding
          response to fluid shear stress
          lymph vessel morphogenesis
          cytoplasmic sequestering of transcription factor
          skin development
          positive regulation of transcription from RNA polymerase II promoter
          digestive tract development
          branching morphogenesis of an epithelial tube
          genitalia development
          detection of mechanical stimulus
          cartilage development
          regulation of mitotic spindle organization
          lung epithelium development
          placenta blood vessel development
          regulation of proteasomal protein catabolic process
          calcium ion transmembrane transport
          renal system development
          mesonephric tubule development
          mesonephric duct development
          metanephric collecting duct development
          metanephric ascending thin limb development
          metanephric proximal tubule development
          metanephric distal tubule morphogenesis
          cation transmembrane transport
          cation channel activity
          calcium channel activity
          protein kinase binding
          protein domain specific binding
          ion channel binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P094541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305421 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094541 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094541 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P094541 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P388040 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64990 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98599 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2806 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101374 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10732132 - -
          Affymetrix Rat Gene 1.0 ST Array 10732130 - -
          Affymetrix Rat Gene 1.0 ST Array 10732115 - -
          Affymetrix Rat Gene 1.0 ST Array 10732154 - -
          Affymetrix Rat Gene 1.0 ST Array 10732116 - -
          Affymetrix Rat Gene 1.0 ST Array 10732142 - -
          Affymetrix Rat Gene 1.0 ST Array 10732157 - -
          Affymetrix Rat Gene 1.0 ST Array 10732146 - -
          Affymetrix Rat Gene 1.0 ST Array 10732153 - -
          Affymetrix Rat Gene 1.0 ST Array 10732117 - -
          Affymetrix Rat Gene 1.0 ST Array 10732134 - -
          Affymetrix Rat Gene 1.0 ST Array 10732122 - -
          Affymetrix Rat Gene 1.0 ST Array 10732128 - -
          Affymetrix Rat Gene 1.0 ST Array 10732141 - -
          Affymetrix Rat Gene 1.0 ST Array 10732118 - -
          Affymetrix Rat Gene 1.0 ST Array 10732125 - -
          Affymetrix Rat Gene 1.0 ST Array 10732148 - -
          Affymetrix Rat Gene 1.0 ST Array 10732137 - -
          Affymetrix Rat Gene 1.0 ST Array 10732119 - -
          Affymetrix Rat Gene 1.0 ST Array 10732114 - -
          Affymetrix Rat Gene 1.0 ST Array 10732136 - -
          Affymetrix Rat Gene 1.0 ST Array 10732131 - -
          Affymetrix Rat Gene 1.0 ST Array 10732133 - -
          Affymetrix Rat Gene 1.0 ST Array 10732145 - -
          Affymetrix Rat Gene 1.0 ST Array 10732159 - -
          Affymetrix Rat Gene 1.0 ST Array 10732149 - -
          Affymetrix Rat Gene 1.0 ST Array 10732150 - -
          Affymetrix Rat Gene 1.0 ST Array 10732140 - -
          Affymetrix Rat Gene 1.0 ST Array 10732120 - -
          Affymetrix Rat Gene 1.0 ST Array 10732144 - -
          Affymetrix Rat Gene 1.0 ST Array 10732129 - -
          Affymetrix Rat Gene 1.0 ST Array 10732121 - -
          Affymetrix Rat Gene 1.0 ST Array 10732135 - -
          Affymetrix Rat Gene 1.0 ST Array 10732158 - -
          Affymetrix Rat Gene 1.0 ST Array 10732138 - -
          Affymetrix Rat Gene 1.0 ST Array 10732155 - -
          Affymetrix Rat Gene 1.0 ST Array 10732143 - -
          Affymetrix Rat Gene 1.0 ST Array 10732123 - -
          Affymetrix Rat Gene 1.0 ST Array 10732156 - -
          Affymetrix Rat Gene 1.0 ST Array 10732126 - -
          Affymetrix Rat Gene 1.0 ST Array 10732147 - -
          Affymetrix Rat Gene 1.0 ST Array 10732152 - -
          Affymetrix Rat Gene 1.0 ST Array 10732124 - -
          Affymetrix Rat Gene 1.0 ST Array 10732139 - -
          Affymetrix Rat Gene 1.0 ST Array 10732127 - -
          Affymetrix Rat Gene 1.0 ST Array 10732151 - -
          Affymetrix Rat Gene 1.1 ST Array 10732113 - -
          Rat Genome 230 2.0 Array 1371029_at - -
          Rat Genome 230 2.0 Array 1371029_PM_at - -
          Affymetrix HT Rat Focus Array 1371029_PM_at - -
          Affymetrix HT Rat Focus Array 1371029_at - -
          Affymetrix Rat Clariom S Assay PSR1000001981.rn.1 TC1000000213.rn.1 RTA TC
          Rat Expression Set 230 1371029_at - -
          Rat Expression Set 230 1371029_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10732113 - -
          Rat Genome U34 Set rc_AA800984_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22268907 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354053 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline