Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01465210_g1
          See other SKP1 GE Assays ›
          Gene Symbol:
          Skp1
          Gene Name:
          S-phase kinase-associated protein 1
          Gene Aliases:
          Skp1a
          Chromosome Location:
          Chr.10: 37594578 - 37609653 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01465210_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Skp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007608.2 NP_001007609.1 4 - 5 400 95
          XM_006246244.2 XP_006246306.1 4 - 5 699 95
          GenBank mRNA BC058152.1 - 4 - 5 403 6890712 95
          FQ210698.1 - 3 - 4 298 95
          FQ212528.1 - 3 - 4 298 95
          FQ212745.1 - 3 - 4 298 95
          FQ212916.1 - 3 - 4 298 95
          FQ213991.1 - 3 - 4 298 95
          FQ214002.1 - 3 - 4 298 95
          FQ214005.1 - 3 - 4 298 95
          FQ214369.1 - 3 - 4 298 95
          FQ214470.1 - 3 - 4 298 95
          FQ215334.1 - 3 - 4 298 95
          FQ215452.1 - 3 - 4 298 95
          FQ219401.1 - 3 - 4 298 95
          FQ224225.1 - 3 - 4 298 95
          FQ224935.1 - 3 - 4 298 95
          FQ229076.1 - 3 - 4 298 95
          FQ229788.1 - 298 95
          FQ230985.1 - 3 - 4 298 95

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Skp1

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ubiquitination
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          histone H2A monoubiquitination
          ubiquitin-protein transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P168112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344720 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111023 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147103 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P168112 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P168112 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P168112 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P168112 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P168103 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27634 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17194 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10733435 - -
          Affymetrix Rat Gene 1.0 ST Array 10733433 - -
          Affymetrix Rat Gene 1.0 ST Array 10733431 - -
          Affymetrix Rat Gene 1.0 ST Array 10733436 - -
          Affymetrix Rat Gene 1.0 ST Array 10733437 - -
          Affymetrix Rat Gene 1.0 ST Array 10733434 - -
          Affymetrix Rat Gene 1.0 ST Array 10733432 - -
          Affymetrix Rat Gene 1.1 ST Array 10733430 - -
          Rat Genome 230 2.0 Array 1372437_PM_at - -
          Rat Genome 230 2.0 Array 1372437_at - -
          Affymetrix HT Rat Focus Array 1372437_PM_at - -
          Affymetrix HT Rat Focus Array 1372437_at - -
          Affymetrix Rat Clariom S Assay PSR1000004789.rn.1 TC1000000569.rn.1 RTA TC
          Rat Expression Set 230 1372437_at - -
          Rat Expression Set 230 1372437_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10733430 - -
          Rat Genome U34 Set rc_AA997522_at - -
          Rat Genome U34 Set rc_AI235972_at - -
          Rat Genome U34 Set rc_AI236635_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22288544 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363654 - -
          RatRef-12 Expression BeadChip ILMN_49450 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline