Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01465407_m1
          See other ST6GALNAC2 GE Assays ›
          Gene Symbol:
          St6galnac2
          Gene Name:
          ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2
          Gene Aliases:
          Siat7b
          Chromosome Location:
          Chr.10: 105650866 - 105668579 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01465407_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: St6galnac2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001031652.1 NP_001026822.1 7 - 8 878 68
          GenBank mRNA AJ634457.1 - 7 - 8 848 68
          BC099812.1 - 7 - 8 878 7929942 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004171.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene St6galnac2

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          sialylation
          sialyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P497302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212461 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132728 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162319 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P497302 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P497302 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P497302 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P497302 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70398 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96249 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33555 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93050 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10749331 - -
          Affymetrix Rat Gene 1.0 ST Array 10749337 - -
          Affymetrix Rat Gene 1.0 ST Array 10749333 - -
          Affymetrix Rat Gene 1.0 ST Array 10749339 - -
          Affymetrix Rat Gene 1.0 ST Array 10749336 - -
          Affymetrix Rat Gene 1.0 ST Array 10749334 - -
          Affymetrix Rat Gene 1.0 ST Array 10749338 - -
          Affymetrix Rat Gene 1.0 ST Array 10749335 - -
          Affymetrix Rat Gene 1.0 ST Array 10749332 - -
          Affymetrix Rat Gene 1.1 ST Array 10749330 - -
          Rat Genome 230 2.0 Array 1379390_at - -
          Rat Genome 230 2.0 Array 1381953_at - -
          Rat Genome 230 2.0 Array 1381953_PM_at - -
          Rat Genome 230 2.0 Array 1378955_at - -
          Rat Genome 230 2.0 Array 1378955_PM_at - -
          Rat Genome 230 2.0 Array 1379390_PM_at - -
          Affymetrix HT Rat Focus Array 1379390_PM_at - -
          Affymetrix HT Rat Focus Array 1379390_at - -
          Affymetrix HT Rat Focus Array 1381953_at - -
          Affymetrix HT Rat Focus Array 1381953_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000038079.rn.1 TC1000004171.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000038078.rn.1 TC1000004171.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000038077.rn.1 TC1000004171.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000038080.rn.1 TC1000004171.rn.1 RTA TC
          Rat Expression Set 230 1379390_PM_at - -
          Rat Expression Set 230 1381953_at - -
          Rat Expression Set 230 1381953_PM_at - -
          Rat Expression Set 230 1378955_at - -
          Rat Expression Set 230 1379390_at - -
          Rat Expression Set 230 1378955_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10749330 - -
          Rat Genome U34 Set rc_AI175990_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351759 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline