Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01467726_g1
          See other TRAF7 GE Assays ›
          Gene Symbol:
          Traf7
          Gene Name:
          TNF receptor-associated factor 7, E3 ubiquitin protein ligase
          Gene Aliases:
          RGD1559653
          Chromosome Location:
          Chr.10: 13874456 - 13893091 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01467726_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Traf7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001127548.1 NP_001121020.1 20 - 21 2098 99
          XM_006245982.2 XP_006246044.1 20 - 21 2192 99
          XM_006245983.2 XP_006246045.1 20 - 21 2187 99
          XM_006245984.2 XP_006246046.1 19 - 20 2033 99
          XM_006245985.2 XP_006246047.1 19 - 20 2010 99
          XM_006245986.2 XP_006246048.1 19 - 20 2030 99
          XM_006245987.2 XP_006246049.1 19 - 20 2007 99
          XM_006245988.2 XP_006246050.1 20 - 21 2229 99
          XM_006245989.2 XP_006246051.1 20 - 21 2226 99
          XM_006245990.2 XP_006246052.1 19 - 20 1979 99
          XM_006245991.2 XP_006246053.1 19 - 20 1983 99
          XM_006245992.2 XP_006246054.1 19 - 20 1976 99
          XM_006245993.2 XP_006246055.1 18 - 19 1969 99
          XM_008767584.1 XP_008765806.1 20 - 21 2245 99
          XM_008767585.1 XP_008765807.1 20 - 21 2242 99
          XM_008767586.1 XP_008765808.1 20 - 21 2240 99
          GenBank mRNA BC161916.1 - 20 - 21 2098 8369808 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002391.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          activation of MAPKKK activity
          apoptotic process
          protein ubiquitination
          regulation of protein localization
          positive regulation of protein sumoylation
          positive regulation of MAPK cascade
          cellular response to interleukin-6
          positive regulation of apoptotic signaling pathway
          ubiquitin-protein transferase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P249391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 485598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643863 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81153 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38401 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P249391 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P249391 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P249391 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P249391 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P479422 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9606 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 510 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10741120 - -
          Affymetrix Rat Gene 1.0 ST Array 10741117 - -
          Affymetrix Rat Gene 1.0 ST Array 10741125 - -
          Affymetrix Rat Gene 1.0 ST Array 10741128 - -
          Affymetrix Rat Gene 1.0 ST Array 10741123 - -
          Affymetrix Rat Gene 1.0 ST Array 10741113 - -
          Affymetrix Rat Gene 1.0 ST Array 10741122 - -
          Affymetrix Rat Gene 1.0 ST Array 10741126 - -
          Affymetrix Rat Gene 1.0 ST Array 10741116 - -
          Affymetrix Rat Gene 1.0 ST Array 10741121 - -
          Affymetrix Rat Gene 1.0 ST Array 10741124 - -
          Affymetrix Rat Gene 1.0 ST Array 10741131 - -
          Affymetrix Rat Gene 1.0 ST Array 10741129 - -
          Affymetrix Rat Gene 1.0 ST Array 10741118 - -
          Affymetrix Rat Gene 1.0 ST Array 10741127 - -
          Affymetrix Rat Gene 1.0 ST Array 10741130 - -
          Affymetrix Rat Gene 1.0 ST Array 10741119 - -
          Affymetrix Rat Gene 1.0 ST Array 10741114 - -
          Affymetrix Rat Gene 1.0 ST Array 10741115 - -
          Affymetrix Rat Gene 1.1 ST Array 10741112 - -
          Rat Genome 230 2.0 Array 1388769_at - -
          Rat Genome 230 2.0 Array 1388769_PM_at - -
          Affymetrix HT Rat Focus Array 1388769_PM_at - -
          Affymetrix HT Rat Focus Array 1388769_at - -
          Affymetrix Rat Clariom S Assay PSR1000021228.rn.1 TC1000002391.rn.1 RTA TC
          Rat Expression Set 230 1388769_PM_at - -
          Rat Expression Set 230 1388769_at - -
          Affymetrix Rat Gene 1.0 ST Array 10741112 - -
          Rat Genome U34 Set rc_AA799600_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20975619 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371994 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline