Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01469304_m1
          See other TTC7A GE Assays ›
          Gene Symbol:
          Ttc7a
          Gene Name:
          tetratricopeptide repeat domain 7A
          Gene Aliases:
          Ttc7
          Chromosome Location:
          Chr.6: 10912383 - 11014279 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01469304_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ttc7a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100756.1 NP_001094226.1 19 - 20 2602 57
          XM_008764464.1 XP_008762686.1 12 - 13 3671 57
          GenBank mRNA BC098811.1 - 4 - 5 559 57
          BC168226.1 - 19 - 20 2602 7371309 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000154.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s)

          cellular iron ion homeostasis
          hemopoiesis

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P100847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638094 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163986 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90675 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157677 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167381 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110383 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127441 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133134 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59916 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26665 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19357 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55008 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165641 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P100847 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P100847 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P100847 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P100847 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9483 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10882785 - -
          Affymetrix Rat Gene 1.0 ST Array 10882797 - -
          Affymetrix Rat Gene 1.0 ST Array 10882788 - -
          Affymetrix Rat Gene 1.0 ST Array 10882800 - -
          Affymetrix Rat Gene 1.0 ST Array 10882798 - -
          Affymetrix Rat Gene 1.0 ST Array 10882794 - -
          Affymetrix Rat Gene 1.0 ST Array 10882786 - -
          Affymetrix Rat Gene 1.0 ST Array 10882799 - -
          Affymetrix Rat Gene 1.0 ST Array 10882781 - -
          Affymetrix Rat Gene 1.0 ST Array 10882793 - -
          Affymetrix Rat Gene 1.0 ST Array 10882790 - -
          Affymetrix Rat Gene 1.0 ST Array 10882796 - -
          Affymetrix Rat Gene 1.0 ST Array 10882801 - -
          Affymetrix Rat Gene 1.0 ST Array 10882795 - -
          Affymetrix Rat Gene 1.0 ST Array 10882780 - -
          Affymetrix Rat Gene 1.0 ST Array 10882782 - -
          Affymetrix Rat Gene 1.0 ST Array 10882791 - -
          Affymetrix Rat Gene 1.0 ST Array 10882789 - -
          Affymetrix Rat Gene 1.0 ST Array 10882787 - -
          Affymetrix Rat Gene 1.0 ST Array 10882792 - -
          Affymetrix Rat Gene 1.0 ST Array 10882784 - -
          Affymetrix Rat Gene 1.0 ST Array 10882783 - -
          Affymetrix Rat Gene 1.1 ST Array 10882777 - -
          Rat Genome 230 2.0 Array 1376974_at - -
          Rat Genome 230 2.0 Array 1376974_PM_at - -
          Affymetrix HT Rat Focus Array 1376974_PM_at - -
          Affymetrix HT Rat Focus Array 1376974_at - -
          Affymetrix Rat Clariom S Assay PSR0600000929.rn.1 TC0600000154.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600000928.rn.1 TC0600000154.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600000930.rn.1 TC0600000154.rn.1 RTA TC
          Rat Expression Set 230 1376974_PM_at - -
          Rat Expression Set 230 1376974_at - -
          Affymetrix Rat Gene 1.0 ST Array 10882777 - -
          Rat Genome U34 Set rc_AA819451_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349781 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline