Search
Search
| Gene Symbol: Haao | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020076.2 | NP_064461.1 | 2 - 3 | 241 | 80 | |
| XM_006239684.2 | XP_006239746.1 | 2 - 3 | 1146 | 80 | ||
| GenBank mRNA | BC085739.1 | - | 2 - 3 | 195 | 7108551 | 80 |
| D44494.1 | - | 2 - 3 | 242 | 80 | ||
| FQ219702.1 | - | 2 - 3 | 255 | 80 | ||
Set Membership: |
|