Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01471804_m1
          See other CLK4 GE Assays ›
          Gene Symbol:
          Clk4
          Gene Name:
          CDC-like kinase 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 36715565 - 36733476 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01471804_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clk4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013041.1 NP_001013059.1 8 - 9 990 104
          XM_006246234.2 XP_006246296.1 9 - 10 1200 104
          XM_006246236.1 XP_006246298.1 12 - 13 1403 104
          XM_006246237.1 XP_006246299.1 10 - 11 1105 104
          XM_006246238.2 XP_006246300.1 9 - 10 2620 104
          XM_008767670.1 XP_008765892.1 10 - 11 2074 104
          XM_008767671.1 XP_008765893.1 9 - 10 1996 104
          XM_008767672.1 XP_008765894.1 9 - 10 1039 104
          XM_008767673.1 XP_008765895.1 8 - 9 2692 104
          XM_008767674.1 XP_008765896.1 7 - 8 2479 104
          XM_008767675.1 XP_008765897.1 10 - 11 1266 104
          XM_008767676.1 XP_008765898.1 8 - 9 991 104
          GenBank mRNA BC079006.1 - 8 - 9 990 7113075 104
          FQ223275.1 - 11 - 12 3050 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000547.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clk4

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-tyrosine phosphorylation
          regulation of RNA splicing
          protein autophosphorylation
          protein serine/threonine kinase activity
          protein tyrosine kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P373209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165797 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741656 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57734 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2111 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P373209 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P373209 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P373209 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P373209 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46020 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10733337 - -
          Affymetrix Rat Gene 1.0 ST Array 10733333 - -
          Affymetrix Rat Gene 1.0 ST Array 10733328 - -
          Affymetrix Rat Gene 1.0 ST Array 10733324 - -
          Affymetrix Rat Gene 1.0 ST Array 10733332 - -
          Affymetrix Rat Gene 1.0 ST Array 10733331 - -
          Affymetrix Rat Gene 1.0 ST Array 10733338 - -
          Affymetrix Rat Gene 1.0 ST Array 10733334 - -
          Affymetrix Rat Gene 1.0 ST Array 10733323 - -
          Affymetrix Rat Gene 1.0 ST Array 10733322 - -
          Affymetrix Rat Gene 1.0 ST Array 10733330 - -
          Affymetrix Rat Gene 1.0 ST Array 10733335 - -
          Affymetrix Rat Gene 1.0 ST Array 10733336 - -
          Affymetrix Rat Gene 1.1 ST Array 10733321 - -
          Rat Genome 230 2.0 Array 1399141_PM_at - -
          Rat Genome 230 2.0 Array 1399141_at - -
          Affymetrix HT Rat Focus Array 1399141_PM_at - -
          Affymetrix HT Rat Focus Array 1399141_at - -
          Affymetrix Rat Clariom S Assay PSR1000004560.rn.1 TC1000000547.rn.1 RTA TC
          Rat Expression Set 230 1399141_at - -
          Rat Expression Set 230 1399141_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10733321 - -
          Rat Genome U34 Set rc_AA924654_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21131943 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371901 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0