Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01472547_m1
          See other ALLC GE Assays ›
          Gene Symbol:
          Allc
          Gene Name:
          allantoicase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 47812989 - 47848070 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01472547_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Allc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013037.1 NP_001013055.1 9 - 10 952 92
          XM_006239944.2 XP_006240006.1 9 - 10 1026 92
          GenBank mRNA BC078971.1 - 9 - 10 952 7111813 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600002246.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Allc

          Gene Ontology Categories:

          Function(s) Process(es)

          allantoin catabolic process
          allantoicase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P454444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662479 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53837 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 2604 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171891 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P454444 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P454444 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P454444 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P454444 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1029408 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40969 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15304 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10889438 - -
          Affymetrix Rat Gene 1.0 ST Array 10889445 - -
          Affymetrix Rat Gene 1.0 ST Array 10889442 - -
          Affymetrix Rat Gene 1.0 ST Array 10889444 - -
          Affymetrix Rat Gene 1.0 ST Array 10889439 - -
          Affymetrix Rat Gene 1.0 ST Array 10889435 - -
          Affymetrix Rat Gene 1.0 ST Array 10889443 - -
          Affymetrix Rat Gene 1.0 ST Array 10889436 - -
          Affymetrix Rat Gene 1.0 ST Array 10889437 - -
          Affymetrix Rat Gene 1.0 ST Array 10889440 - -
          Affymetrix Rat Gene 1.0 ST Array 10889441 - -
          Affymetrix Rat Gene 1.1 ST Array 10889434 - -
          Affymetrix Rat Clariom S Assay PSR0600016197.rn.1 TC0600002246.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10889434 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22308813 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356426 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline