Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01474607_g1
          See other FAM162A GE Assays ›
          Gene Symbol:
          Fam162a
          Gene Name:
          family with sequence similarity 162, member A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 67618652 - 67646253 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01474607_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam162a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001029903.1 NP_001025074.1 3 - 4 258 65
          XM_006248413.2 XP_006248475.1 3 - 4 346 65
          GenBank mRNA BC097971.1 - 3 - 4 292 7442058 65
          M17412.1 - 2 - 3 231 65

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam162a

          Gene Ontology Categories:

          Function(s)

          activation of cysteine-type endopeptidase activity involved in apoptotic process
          obsolete transformed cell apoptotic process
          positive regulation of apoptotic process
          neuron apoptotic process
          cellular response to hypoxia
          positive regulation of release of cytochrome c from mitochondria

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P002770 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P062760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 100938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903052 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11645 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89061 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116874 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11507 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159631 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P002770 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062760 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P002770 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P062760 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P002770 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P062760 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P367767 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700230337 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700435991 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23682 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66119 - -
          Agilent Rat cDNA Microarray (G4105A) 700230337 - -
          Agilent Rat cDNA Microarray (G4105A) 700435991 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10751458 - -
          Affymetrix Rat Gene 1.0 ST Array 10751451 - -
          Affymetrix Rat Gene 1.0 ST Array 10751454 - -
          Affymetrix Rat Gene 1.0 ST Array 10751455 - -
          Affymetrix Rat Gene 1.0 ST Array 10751456 - -
          Affymetrix Rat Gene 1.0 ST Array 10751452 - -
          Affymetrix Rat Gene 1.0 ST Array 10751453 - -
          Affymetrix Rat Gene 1.1 ST Array 10751450 - -
          Affymetrix Rat Clariom S Assay PSR1100003742.rn.1 TC1100000693.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100003753.rn.1 TC1100000693.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10751450 - -
          Rat Genome U34 Set M17412_at - -
          Rat Genome U34 Set rc_AI234915_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21197954 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline