Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01476554_m1
          See other SMG9 GE Assays ›
          Gene Symbol:
          Smg9
          Gene Name:
          SMG9 nonsense mediated mRNA decay factor
          Gene Aliases:
          RGD1309888
          Chromosome Location:
          Chr.1: 81259447 - 81282893 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01476554_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smg9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014243.1 NP_001014265.1 13 - 14 1559 88
          XM_006228450.2 XP_006228512.1 13 - 14 2490 88
          GenBank mRNA BC087051.1 - 13 - 14 1559 7115791 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001039.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smg9

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P375042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907039 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66130 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76420 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69509 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P375042 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P375042 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P375042 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P375042 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80847 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63443 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10704951 - -
          Affymetrix Rat Gene 1.0 ST Array 10704948 - -
          Affymetrix Rat Gene 1.0 ST Array 10704954 - -
          Affymetrix Rat Gene 1.0 ST Array 10704952 - -
          Affymetrix Rat Gene 1.0 ST Array 10704949 - -
          Affymetrix Rat Gene 1.0 ST Array 10704944 - -
          Affymetrix Rat Gene 1.0 ST Array 10704953 - -
          Affymetrix Rat Gene 1.0 ST Array 10704946 - -
          Affymetrix Rat Gene 1.0 ST Array 10704942 - -
          Affymetrix Rat Gene 1.0 ST Array 10704943 - -
          Affymetrix Rat Gene 1.0 ST Array 10704947 - -
          Affymetrix Rat Gene 1.0 ST Array 10704945 - -
          Affymetrix Rat Gene 1.0 ST Array 10704955 - -
          Affymetrix Rat Gene 1.0 ST Array 10704950 - -
          Affymetrix Rat Gene 1.1 ST Array 10704941 - -
          Rat Genome 230 2.0 Array 1373655_PM_at - -
          Rat Genome 230 2.0 Array 1373655_at - -
          Affymetrix HT Rat Focus Array 1373655_PM_at - -
          Affymetrix HT Rat Focus Array 1373655_at - -
          Affymetrix Rat Clariom S Assay PSR0100007328.rn.1 TC0100001039.rn.1 RTA TC
          Rat Expression Set 230 1373655_at - -
          Rat Expression Set 230 1373655_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10704941 - -
          Rat Genome U34 Set rc_AA925547_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21542752 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361479 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline