Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01477341_m1
          See other LIPI GE Assays ›
          Gene Symbol:
          Lipi
          Gene Name:
          lipase, member I
          Gene Aliases:
          Liph
          Chromosome Location:
          Chr.11: 13960246 - 13999869 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01477341_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lipi
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001105899.1 NP_001099369.1 3 - 4 589 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001140.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          lipase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          lipid catabolic process
          molecular_function
          phospholipase activity
          heparin binding
          carboxylic ester hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P040371 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P097302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 890877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731189 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84704 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56387 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81071 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097302 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040371 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097302 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040371 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P097302 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P040371 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P499622 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60924 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10752731 - -
          Affymetrix Rat Gene 1.0 ST Array 10752730 - -
          Affymetrix Rat Gene 1.0 ST Array 10752729 - -
          Affymetrix Rat Gene 1.0 ST Array 10752734 - -
          Affymetrix Rat Gene 1.0 ST Array 10752735 - -
          Affymetrix Rat Gene 1.0 ST Array 10752737 - -
          Affymetrix Rat Gene 1.0 ST Array 10752733 - -
          Affymetrix Rat Gene 1.0 ST Array 10752736 - -
          Affymetrix Rat Gene 1.0 ST Array 10752732 - -
          Affymetrix Rat Gene 1.1 ST Array 10752728 - -
          Affymetrix Rat Clariom S Assay PSR1100006655.rn.1 TC1100001140.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100006650.rn.1 TC1100001140.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100006652.rn.1 TC1100001140.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100006648.rn.1 TC1100001140.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100006649.rn.1 TC1100001140.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100006653.rn.1 TC1100001140.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100006651.rn.1 TC1100001140.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100006654.rn.1 TC1100001140.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10752728 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21985126 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361597 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline