Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01478843_m1
          See other SCGB2A2 GE Assays ›
          Gene Symbol:
          Scgb2a2
          Gene Name:
          secretoglobin, family 2A, member 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 225899014 - 225902676 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01478843_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scgb2a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_203333.2 NP_976078.1 1 - 2 144 157
          XM_008760232.1 XP_008758454.1 1 - 2 443 157
          GenBank mRNA BC062401.1 - 1 - 2 104 5621455 157
          CB791758.1 - 1 - 2 156 157
          J00772.1 - 1 - 2 95 157

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003175.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scgb2a2

          Gene Ontology Categories:

          Function(s) Process(es)

          protein tetramerization
          steroid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P105063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548362 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133555 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105063 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P105063 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P105063 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P510425 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91817 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713820 - -
          Affymetrix Rat Gene 1.0 ST Array 10713821 - -
          Affymetrix Rat Gene 1.0 ST Array 10713812 - -
          Affymetrix Rat Gene 1.0 ST Array 10713822 - -
          Affymetrix Rat Gene 1.0 ST Array 10713814 - -
          Affymetrix Rat Gene 1.0 ST Array 10713819 - -
          Affymetrix Rat Gene 1.0 ST Array 10713813 - -
          Affymetrix Rat Gene 1.0 ST Array 10713815 - -
          Affymetrix Rat Gene 1.1 ST Array 10713818 - -
          Affymetrix Rat Gene 1.1 ST Array 10713811 - -
          Rat Genome 230 2.0 Array 1367554_at - -
          Rat Genome 230 2.0 Array 1367554_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100026225.rn.1 TC0100003175.rn.1 RTA TC
          Rat Expression Set 230 1367554_PM_at - -
          Rat Expression Set 230 1367554_at - -
          Affymetrix Rat Gene 1.0 ST Array 10713811 - -
          Affymetrix Rat Gene 1.0 ST Array 10713818 - -
          Rat Genome U34 Set J00772_s_at - -
          Rat Genome U34 Set J00777_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21819715 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355673 - -
          RatRef-12 Expression BeadChip ILMN_53523 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0