Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01481712_m1
          See other NCOR2 GE Assays ›
          Gene Symbol:
          Ncor2
          Gene Name:
          nuclear receptor co-repressor 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 36871909 - 37033701 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01481712_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ncor2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108334.1 NP_001101804.1 34 - 35 5271 67
          XM_006249305.2 XP_006249367.1 36 - 37 5584 67
          XM_006249306.2 XP_006249368.1 36 - 37 5581 67
          XM_006249307.2 XP_006249369.1 36 - 37 5581 67
          XM_006249309.2 XP_006249371.1 36 - 37 5581 67
          XM_006249311.2 XP_006249373.1 36 - 37 5584 67
          XM_006249312.2 XP_006249374.1 36 - 37 5527 67
          XM_006249314.2 XP_006249376.1 36 - 37 5584 67
          XM_006249315.2 XP_006249377.1 36 - 37 5584 67
          XM_006249317.2 XP_006249379.1 36 - 37 5527 67
          XM_006249319.2 XP_006249381.1 36 - 37 5527 67
          XM_006249320.2 XP_006249382.1 36 - 37 5521 67
          XM_006249321.2 XP_006249383.1 36 - 37 5584 67
          XM_006249322.2 XP_006249384.1 36 - 37 5584 67
          XM_006249323.2 XP_006249385.1 36 - 37 5521 67
          XM_008769236.1 XP_008767458.1 36 - 37 5566 67
          XM_008769237.1 XP_008767459.1 36 - 37 5509 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000582.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          heart morphogenesis
          lactation
          response to organonitrogen compound
          telencephalon development
          cell proliferation in forebrain
          forebrain development
          response to estradiol
          glucose homeostasis
          estrous cycle
          negative regulation of fat cell differentiation
          negative regulation of transcription, DNA-templated
          white fat cell differentiation
          Type I pneumocyte differentiation
          regulation of cellular ketone metabolic process by negative regulation of transcription from RNA polymerase II promoter
          positive regulation of protein deacetylation
          RNA polymerase II regulatory region DNA binding
          DNA binding
          chromatin binding
          transcription corepressor activity
          Notch binding
          protein binding
          ligand-dependent nuclear receptor binding
          protein complex binding
          glucocorticoid receptor binding
          histone deacetylase binding
          retinoic acid receptor binding
          sequence-specific DNA binding
          retinoid X receptor binding
          protein N-terminus binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR1200004942.rn.1 TC1200000582.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline