Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01484390_m1
          See other NDUFS3 GE Assays ›
          Gene Symbol:
          Ndufs3
          Gene Name:
          NADH dehydrogenase (ubiquinone) Fe-S protein 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 79721686 - 79728863 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01484390_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufs3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106489.1 NP_001099959.1 4 - 5 554 62
          GenBank mRNA FQ219879.1 - 4 - 5 381 62
          FQ229748.1 - 4 - 5 401 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003642.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          substantia nigra development
          negative regulation of cell growth
          oxidation-reduction process
          reactive oxygen species metabolic process
          negative regulation of intrinsic apoptotic signaling pathway
          molecular_function
          NADH dehydrogenase activity
          NADH dehydrogenase (ubiquinone) activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P702941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567198 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4148 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P702941 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P702941 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P702941 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P702941 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10847289 - -
          Affymetrix Rat Gene 1.0 ST Array 10847290 - -
          Affymetrix Rat Gene 1.0 ST Array 10847288 - -
          Affymetrix Rat Gene 1.0 ST Array 10847285 - -
          Affymetrix Rat Gene 1.0 ST Array 10847284 - -
          Affymetrix Rat Gene 1.0 ST Array 10847287 - -
          Affymetrix Rat Gene 1.0 ST Array 10847286 - -
          Affymetrix Rat Gene 1.1 ST Array 10847283 - -
          Rat Genome 230 2.0 Array 1388364_PM_at - -
          Rat Genome 230 2.0 Array 1388364_at - -
          Affymetrix HT Rat Focus Array 1388364_at - -
          Affymetrix HT Rat Focus Array 1388364_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300027608.rn.1 TC0300003642.rn.1 RTA TC
          Rat Expression Set 230 1388364_PM_at - -
          Rat Expression Set 230 1388364_at - -
          Affymetrix Rat Gene 1.0 ST Array 10847283 - -
          Rat Genome U34 Set rc_AI229680_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22176396 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361813 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline