Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01487445_m1
          See other RBM17 GE Assays ›
          Gene Symbol:
          Rbm17
          Gene Name:
          RNA binding motif protein 17
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 70586343 - 70603289 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01487445_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbm17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013058.1 NP_001013076.1 6 - 7 694 94
          XM_006254205.2 XP_006254267.1 6 - 7 801 94
          GenBank mRNA BC079248.1 - 6 - 7 694 7132972 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000923.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbm17

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          alternative mRNA splicing, via spliceosome
          nucleotide binding
          nucleic acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74039 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148980 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26628 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11313 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11313 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11313 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P368101 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11313 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77919 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5118 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10796137 - -
          Affymetrix Rat Gene 1.0 ST Array 10796143 - -
          Affymetrix Rat Gene 1.0 ST Array 10796142 - -
          Affymetrix Rat Gene 1.0 ST Array 10796146 - -
          Affymetrix Rat Gene 1.0 ST Array 10796140 - -
          Affymetrix Rat Gene 1.0 ST Array 10796136 - -
          Affymetrix Rat Gene 1.0 ST Array 10796135 - -
          Affymetrix Rat Gene 1.0 ST Array 10796141 - -
          Affymetrix Rat Gene 1.0 ST Array 10796144 - -
          Affymetrix Rat Gene 1.0 ST Array 10796139 - -
          Affymetrix Rat Gene 1.0 ST Array 10796138 - -
          Affymetrix Rat Gene 1.0 ST Array 10796145 - -
          Affymetrix Rat Gene 1.1 ST Array 10796134 - -
          Rat Genome 230 2.0 Array 1388355_PM_at - -
          Rat Genome 230 2.0 Array 1388355_at - -
          Rat Genome 230 2.0 Array 1396401_at - -
          Rat Genome 230 2.0 Array 1396401_PM_at - -
          Rat Genome 230 2.0 Array 1396185_at - -
          Rat Genome 230 2.0 Array 1396185_PM_at - -
          Affymetrix HT Rat Focus Array 1388355_at - -
          Affymetrix HT Rat Focus Array 1396401_PM_at - -
          Affymetrix HT Rat Focus Array 1396401_at - -
          Affymetrix HT Rat Focus Array 1396185_PM_at - -
          Affymetrix HT Rat Focus Array 1396185_at - -
          Affymetrix HT Rat Focus Array 1388355_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700006378.rn.1 TC1700000923.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700006376.rn.1 TC1700000923.rn.1 RTA TC
          Rat Expression Set 230 1388355_PM_at - -
          Rat Expression Set 230 1388355_at - -
          Rat Expression Set 230 1396401_at - -
          Rat Expression Set 230 1396185_PM_at - -
          Rat Expression Set 230 1396185_at - -
          Rat Expression Set 230 1396401_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10796134 - -
          Rat Genome U34 Set rc_AI072014_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21920192 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358042 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline