Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01493919_m1
          See other TRAPPC13 GE Assays ›
          Gene Symbol:
          Trappc13
          Gene Name:
          trafficking protein particle complex 13
          Gene Aliases:
          RGD1306583
          Chromosome Location:
          Chr.2: 34223396 - 34254988 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01493919_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trappc13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013908.2 NP_001013930.2 10 - 11 1173 128
          XM_006231884.2 XP_006231946.1 10 - 11 1212 128
          XM_006231885.2 XP_006231947.1 9 - 10 1194 128
          XM_006231886.2 XP_006231948.1 9 - 10 1194 128
          GenBank mRNA BC088172.1 - 10 - 11 1147 7323103 128
          CO397574.1 - 4 - 5 424 128
          FQ217190.1 - 1156 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002790.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trappc13

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P815485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158441 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841996 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141155 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128698 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135020 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P815485 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P815485 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P815485 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P815485 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1044244 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32736 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1087 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10821141 - -
          Affymetrix Rat Gene 1.0 ST Array 10821145 - -
          Affymetrix Rat Gene 1.0 ST Array 10821142 - -
          Affymetrix Rat Gene 1.0 ST Array 10821140 - -
          Affymetrix Rat Gene 1.0 ST Array 10821138 - -
          Affymetrix Rat Gene 1.0 ST Array 10821135 - -
          Affymetrix Rat Gene 1.0 ST Array 10821143 - -
          Affymetrix Rat Gene 1.0 ST Array 10821136 - -
          Affymetrix Rat Gene 1.0 ST Array 10821137 - -
          Affymetrix Rat Gene 1.0 ST Array 10821139 - -
          Affymetrix Rat Gene 1.0 ST Array 10821146 - -
          Affymetrix Rat Gene 1.0 ST Array 10821144 - -
          Affymetrix Rat Gene 1.1 ST Array 10821134 - -
          Rat Genome 230 2.0 Array 1379795_PM_at - -
          Rat Genome 230 2.0 Array 1372547_PM_at - -
          Rat Genome 230 2.0 Array 1372547_at - -
          Rat Genome 230 2.0 Array 1379795_at - -
          Affymetrix HT Rat Focus Array 1372547_at - -
          Affymetrix HT Rat Focus Array 1379795_PM_at - -
          Affymetrix HT Rat Focus Array 1372547_PM_at - -
          Affymetrix HT Rat Focus Array 1379795_at - -
          Affymetrix Rat Clariom S Assay PSR0200019798.rn.1 TC0200002790.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200019800.rn.1 TC0200002790.rn.1 RTA TC
          Rat Expression Set 230 1372547_PM_at - -
          Rat Expression Set 230 1372547_at - -
          Rat Expression Set 230 1379795_PM_at - -
          Rat Expression Set 230 1379795_at - -
          Affymetrix Rat Gene 1.0 ST Array 10821134 - -
          Rat Genome U34 Set rc_AA819224_at - -
          Rat Genome U34 Set rc_AI175034_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22109495 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358860 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline