Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01494554_m1
          See other GRAMD3 GE Assays ›
          Gene Symbol:
          Gramd3
          Gene Name:
          GRAM domain containing 3
          Gene Aliases:
          RGD1311016
          Chromosome Location:
          Chr.18: 51492196 - 51591028 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01494554_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gramd3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014011.1 NP_001014033.1 12 - 13 1239 95
          XM_006254748.2 XP_006254810.1 12 - 13 1341 95
          XM_008772140.1 XP_008770362.1 12 - 13 1166 95
          GenBank mRNA BC090001.1 - 12 - 13 1239 7321780 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000549.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gramd3

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P026818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87525 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172964 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33793 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70272 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161291 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55051 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35608 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40701 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128149 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103328 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85765 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P026818 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P026818 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P026818 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P175681 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1687 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29950 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104243 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54587 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64161 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51012 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95758 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98796 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82195 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10801803 - -
          Affymetrix Rat Gene 1.0 ST Array 10801805 - -
          Affymetrix Rat Gene 1.0 ST Array 10801801 - -
          Affymetrix Rat Gene 1.0 ST Array 10801808 - -
          Affymetrix Rat Gene 1.0 ST Array 10801798 - -
          Affymetrix Rat Gene 1.0 ST Array 10801800 - -
          Affymetrix Rat Gene 1.0 ST Array 10801797 - -
          Affymetrix Rat Gene 1.0 ST Array 10801809 - -
          Affymetrix Rat Gene 1.0 ST Array 10801795 - -
          Affymetrix Rat Gene 1.0 ST Array 10801804 - -
          Affymetrix Rat Gene 1.0 ST Array 10801806 - -
          Affymetrix Rat Gene 1.0 ST Array 10801799 - -
          Affymetrix Rat Gene 1.0 ST Array 10801807 - -
          Affymetrix Rat Gene 1.0 ST Array 10801802 - -
          Affymetrix Rat Gene 1.1 ST Array 10801794 - -
          Rat Genome 230 2.0 Array 1384177_PM_at - -
          Rat Genome 230 2.0 Array 1398664_PM_at - -
          Rat Genome 230 2.0 Array 1374043_at - -
          Rat Genome 230 2.0 Array 1398664_at - -
          Rat Genome 230 2.0 Array 1384177_at - -
          Rat Genome 230 2.0 Array 1374043_PM_at - -
          Affymetrix HT Rat Focus Array 1374043_at - -
          Affymetrix HT Rat Focus Array 1384177_PM_at - -
          Affymetrix HT Rat Focus Array 1398664_at - -
          Affymetrix HT Rat Focus Array 1398664_PM_at - -
          Affymetrix HT Rat Focus Array 1374043_PM_at - -
          Affymetrix HT Rat Focus Array 1384177_at - -
          Affymetrix Rat Clariom S Assay PSR1800003984.rn.1 TC1800000549.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800003983.rn.1 TC1800000549.rn.1 RTA TC
          Rat Expression Set 230 1374043_at - -
          Rat Expression Set 230 1374043_PM_at - -
          Rat Expression Set 230 1384177_at - -
          Rat Expression Set 230 1398664_PM_at - -
          Rat Expression Set 230 1398664_at - -
          Rat Expression Set 230 1384177_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10801794 - -
          Rat Genome U34 Set rc_AA850738_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22404153 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1651116 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline