Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01495731_m1
          See other P4HA2 GE Assays ›
          Gene Symbol:
          P4ha2
          Gene Name:
          prolyl 4-hydroxylase, alpha polypeptide II
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 39435227 - 39464134 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01495731_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: P4ha2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108275.1 NP_001101745.1 3 - 4 347 69
          XM_006246300.1 XP_006246362.1 3 - 4 316 69
          XM_006246302.1 XP_006246364.1 3 - 4 316 69
          XM_008767691.1 XP_008765913.1 3 - 4 344 69
          GenBank mRNA FQ228519.1 - 3 - 4 252 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000595.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-proline hydroxylation to 4-hydroxy-L-proline
          oxidation-reduction process
          procollagen-proline 4-dioxygenase activity
          iron ion binding
          oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
          L-ascorbic acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P032087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967844 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160369 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164203 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59170 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032087 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P032087 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P032087 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P445938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 760 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10733569 - -
          Affymetrix Rat Gene 1.0 ST Array 10733566 - -
          Affymetrix Rat Gene 1.0 ST Array 10733570 - -
          Affymetrix Rat Gene 1.0 ST Array 10733581 - -
          Affymetrix Rat Gene 1.0 ST Array 10733578 - -
          Affymetrix Rat Gene 1.0 ST Array 10733568 - -
          Affymetrix Rat Gene 1.0 ST Array 10733580 - -
          Affymetrix Rat Gene 1.0 ST Array 10733579 - -
          Affymetrix Rat Gene 1.0 ST Array 10733577 - -
          Affymetrix Rat Gene 1.0 ST Array 10733565 - -
          Affymetrix Rat Gene 1.0 ST Array 10733571 - -
          Affymetrix Rat Gene 1.0 ST Array 10733575 - -
          Affymetrix Rat Gene 1.0 ST Array 10733574 - -
          Affymetrix Rat Gene 1.0 ST Array 10733567 - -
          Affymetrix Rat Gene 1.0 ST Array 10733572 - -
          Affymetrix Rat Gene 1.0 ST Array 10733573 - -
          Affymetrix Rat Gene 1.1 ST Array 10733564 - -
          Rat Genome 230 2.0 Array 1372610_PM_at - -
          Rat Genome 230 2.0 Array 1372610_at - -
          Rat Genome 230 2.0 Array 1375332_PM_at - -
          Rat Genome 230 2.0 Array 1375332_at - -
          Affymetrix HT Rat Focus Array 1372610_PM_at - -
          Affymetrix HT Rat Focus Array 1372610_at - -
          Affymetrix HT Rat Focus Array 1375332_PM_at - -
          Affymetrix HT Rat Focus Array 1375332_at - -
          Affymetrix Rat Clariom S Assay PSR1000005073.rn.1 TC1000000595.rn.1 RTA TC
          Rat Expression Set 230 1375332_PM_at - -
          Rat Expression Set 230 1372610_at - -
          Rat Expression Set 230 1375332_at - -
          Rat Expression Set 230 1372610_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10733564 - -
          Rat Genome U34 Set rc_AI175454_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20879991 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353974 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline