Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rn01498580_m1
See other IGFL3 GE Assays ›
Gene Symbol:
Igfl3
Gene Name:
IGF-like family member 3
Gene Aliases:
Ceacam12
Chromosome Location:
Chr.1: 79474809 - 79481865 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID
Rn01498580_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Igfl3
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001108474.1
NP_001101944.1
3 - 4
900
101
GenBank mRNA
BC158846.1
-
3 - 4
885
7443540
101
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0100000962.rn.1
Set Membership:
Amplicon length greater than or equal to 101
3' Most
Mammalian Gene Collection
Recommended Assay
Rattus norvegicus
1700
Probe spans exons
Panther Classification:
Molecular Function -
immunoglobulin receptor superfamily
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name
Probe IDs
Targets
Target Type
Agilent Whole Rat Genome Microarray 4x44K v2
A_64_P164632
-
-
Agilent SurePrint G3 Rat CGH Microarray 1x1M
122947
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P164632
-
-
Agilent Whole Rat Genome Microarray 4x44K v3
A_64_P164632
-
-
Agilent SurePrint G3 Rat GE 8x60K Microarray
A_64_P164632
-
-
SurePrint G3 Rat Gene Expression Microarray G4131A
A_44_P344543
-
-
Array Name
Probe IDs
Targets
Target Type
Rat Genome 230 2.0 Array
1394892_at
-
-
Rat Genome 230 2.0 Array
1373342_at
-
-
Rat Genome 230 2.0 Array
1373342_PM_at
-
-
Rat Genome 230 2.0 Array
1394892_PM_at
-
-
Affymetrix HT Rat Focus Array
1373342_PM_at
-
-
Affymetrix HT Rat Focus Array
1373342_at
-
-
Affymetrix Rat Clariom S Assay
PSR0100006691.rn.1
TC0100000962.rn.1
RTA TC
Affymetrix Rat Clariom S Assay
PSR0100006694.rn.1
TC0100000962.rn.1
RTA TC
Rat Expression Set 230
1373342_at
-
-
Rat Expression Set 230
1373342_PM_at
-
-
Rat Expression Set 230
1394892_PM_at
-
-
Rat Expression Set 230
1394892_at
-
-
Rat Genome U34 Set
rc_AI639397_s_at
-
-
Rat Genome U34 Set
rc_AI013253_i_at
-
-
Array Name
Probe IDs
Targets
Target Type
ABI Rat Genome Survey Microarray
20701680
-
-
Array Name
Probe IDs
Targets
Target Type
Illumina ratRef-12 v1.0 expression beadchip
ILMN_1357665
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline