Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01502203_m1
          See other CYP2C11 GE Assays ›
          Gene Symbol:
          Cyp2c11
          Gene Name:
          cytochrome P450, subfamily 2, polypeptide 11
          Gene Aliases:
          CYP2CII, Cyp2c
          Chromosome Location:
          Chr.1: 257676172 - 258004428 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01502203_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp2c11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019184.2 NP_062057.2 7 - 8 1158 103
          XM_008760364.1 XP_008758586.1 6 - 7 986 103
          GenBank mRNA BC088146.1 - 7 - 8 1148 7369647 103
          J02657.1 - 7 - 8 1158 103
          U33173.1 - 7 - 8 1158 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003579.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp2c11

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          epoxygenase P450 pathway
          exogenous drug catabolic process
          oxidation-reduction process
          iron ion binding
          testosterone 16-alpha-hydroxylase activity
          arachidonic acid epoxygenase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          heme binding
          aromatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P280786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916129 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63691 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102181 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103751 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P280786 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P280786 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P280786 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P280786 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58883 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79703 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10715219 - -
          Affymetrix Rat Gene 1.0 ST Array 10715216 - -
          Affymetrix Rat Gene 1.0 ST Array 10715213 - -
          Affymetrix Rat Gene 1.0 ST Array 10715217 - -
          Affymetrix Rat Gene 1.0 ST Array 10715220 - -
          Affymetrix Rat Gene 1.0 ST Array 10715218 - -
          Affymetrix Rat Gene 1.0 ST Array 10715212 - -
          Affymetrix Rat Gene 1.0 ST Array 10715215 - -
          Affymetrix Rat Gene 1.0 ST Array 10715221 - -
          Affymetrix Rat Gene 1.0 ST Array 10715214 - -
          Affymetrix Rat Gene 1.1 ST Array 10715211 - -
          Rat Genome 230 2.0 Array 1387328_PM_at - -
          Rat Genome 230 2.0 Array 1387328_at - -
          Affymetrix HT Rat Focus Array 1387328_PM_at - -
          Affymetrix HT Rat Focus Array 1387328_at - -
          Affymetrix Rat Clariom S Assay PSR0100029073.rn.1 TC0100003579.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029075.rn.1 TC0100003579.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029072.rn.1 TC0100003579.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029071.rn.1 TC0100003579.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100029074.rn.1 TC0100003579.rn.1 RTA TC
          Rat Expression Set 230 1387328_at - -
          Rat Expression Set 230 1387328_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10715211 - -
          Rat Genome U34 Set M18363cds_s_at - -
          Rat Genome U34 Set X79081mRNA_f_at - -
          Rat Genome U34 Set J02657_s_at - -
          Rat Genome U34 Set M18363cds_s_at - -
          Rat Genome U34 Set J02657_s_at - -
          Rat Genome U34 Set X79081mRNA_f_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20944728 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357969 - -
          RatRef-12 Expression BeadChip ILMN_60675 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline