Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01502760_m1
          See other CPSF6 GE Assays ›
          Gene Symbol:
          Cpsf6
          Gene Name:
          cleavage and polyadenylation specific factor 6, 68kDa
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 60385688 - 60417552 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01502760_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cpsf6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106785.1 NP_001100255.1 7 - 8 1355 118
          XM_006241372.1 XP_006241434.1 8 - 9 1498 118
          XM_006241373.1 XP_006241435.1 8 - 9 1498 118
          XM_006241374.1 XP_006241436.1 8 - 9 1498 118
          XM_008765386.1 XP_008763608.1 8 - 9 1498 118
          XM_008765387.1 XP_008763609.1 7 - 8 1387 118
          XM_008765388.1 XP_008763610.1 7 - 8 1387 118
          XM_008765390.1 XP_008763612.1 8 - 9 1423 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002756.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA processing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA polyadenylation
          mRNA processing
          biological_process
          protein tetramerization
          nucleotide binding
          molecular_function
          RNA binding
          mRNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P501827 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P874636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235298 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107256 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40609 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125154 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48577 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P501827 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P874636 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P874636 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P501827 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P501827 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P874636 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1040889 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P501827 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P683666 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34466 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10902562 - -
          Affymetrix Rat Gene 1.0 ST Array 10902556 - -
          Affymetrix Rat Gene 1.0 ST Array 10902553 - -
          Affymetrix Rat Gene 1.0 ST Array 10902554 - -
          Affymetrix Rat Gene 1.0 ST Array 10902559 - -
          Affymetrix Rat Gene 1.0 ST Array 10902555 - -
          Affymetrix Rat Gene 1.0 ST Array 10902560 - -
          Affymetrix Rat Gene 1.0 ST Array 10902558 - -
          Affymetrix Rat Gene 1.0 ST Array 10902561 - -
          Affymetrix Rat Gene 1.1 ST Array 10902552 - -
          Rat Genome 230 2.0 Array 1383504_at - -
          Rat Genome 230 2.0 Array 1395425_PM_at - -
          Rat Genome 230 2.0 Array 1379729_at - -
          Rat Genome 230 2.0 Array 1395082_PM_at - -
          Rat Genome 230 2.0 Array 1395425_at - -
          Rat Genome 230 2.0 Array 1379729_PM_at - -
          Rat Genome 230 2.0 Array 1395082_at - -
          Rat Genome 230 2.0 Array 1383504_PM_at - -
          Rat Genome 230 2.0 Array 1376811_PM_a_at - -
          Rat Genome 230 2.0 Array 1376811_a_at - -
          Affymetrix HT Rat Focus Array 1383504_at - -
          Affymetrix HT Rat Focus Array 1376811_PM_a_at - -
          Affymetrix HT Rat Focus Array 1383504_PM_at - -
          Affymetrix HT Rat Focus Array 1376811_a_at - -
          Affymetrix HT Rat Focus Array 1379729_PM_at - -
          Affymetrix HT Rat Focus Array 1379729_at - -
          Affymetrix Rat Clariom S Assay PSR0700021464.rn.1 TC0700002756.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700021459.rn.1 TC0700002756.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700021460.rn.1 TC0700002756.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700021457.rn.1 TC0700002756.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700021466.rn.1 TC0700002756.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700021456.rn.1 TC0700002756.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700021452.rn.1 TC0700002756.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700021453.rn.1 TC0700002756.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700021454.rn.1 TC0700002756.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700021455.rn.1 TC0700002756.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700021458.rn.1 TC0700002756.rn.1 RTA TC
          Rat Expression Set 230 1376811_PM_a_at - -
          Rat Expression Set 230 1376811_a_at - -
          Rat Expression Set 230 1383504_at - -
          Rat Expression Set 230 1395082_PM_at - -
          Rat Expression Set 230 1379729_PM_at - -
          Rat Expression Set 230 1395425_at - -
          Rat Expression Set 230 1383504_PM_at - -
          Rat Expression Set 230 1395082_at - -
          Rat Expression Set 230 1395425_PM_at - -
          Rat Expression Set 230 1379729_at - -
          Affymetrix Rat Gene 1.0 ST Array 10902552 - -
          Rat Genome U34 Set rc_AI071906_at - -
          Rat Genome U34 Set AA850319_at - -
          Rat Genome U34 Set rc_AI169591_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366756 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline