Search
Search
| Gene Symbol: Tmem19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199098.1 | NP_954529.1 | 4 - 5 | 928 | 68 | |
| XM_006241347.2 | XP_006241409.1 | 5 - 6 | 1025 | 68 | ||
| XM_006241348.2 | XP_006241410.1 | 5 - 6 | 878 | 68 | ||
| XM_006241349.1 | XP_006241411.1 | 5 - 6 | 763 | 68 | ||
| XM_008765355.1 | XP_008763577.1 | 4 - 5 | 974 | 68 | ||
| GenBank mRNA | BC061871.1 | - | 4 - 5 | 928 | 5598899 | 68 |
Set Membership: |
|