Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01503703_m1
          See other MDM1 GE Assays ›
          Gene Symbol:
          Mdm1
          Gene Name:
          Mdm1 nuclear protein homolog (mouse)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 61165643 - 61200865 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01503703_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mdm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017459.2 NP_001017459.2 14 - 15 2175 100
          XM_006241385.2 XP_006241447.1 14 - 15 2209 100
          XM_006241387.2 XP_006241449.1 14 - 15 2199 100
          XM_006241388.2 XP_006241450.1 13 - 14 2182 100
          XM_008765400.1 XP_008763622.1 16 - 17 2489 100
          GenBank mRNA BC087101.1 - 14 - 15 2141 7192579 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000771.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mdm1

          Gene Ontology Categories:

          Function(s)

          retina development in camera-type eye

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P129618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710954 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109843 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119387 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95597 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129618 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P129618 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P129618 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P248598 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103326 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64005 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80718 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10895614 - -
          Affymetrix Rat Gene 1.0 ST Array 10895609 - -
          Affymetrix Rat Gene 1.0 ST Array 10895615 - -
          Affymetrix Rat Gene 1.0 ST Array 10895603 - -
          Affymetrix Rat Gene 1.0 ST Array 10895602 - -
          Affymetrix Rat Gene 1.0 ST Array 10895612 - -
          Affymetrix Rat Gene 1.0 ST Array 10895606 - -
          Affymetrix Rat Gene 1.0 ST Array 10895605 - -
          Affymetrix Rat Gene 1.0 ST Array 10895608 - -
          Affymetrix Rat Gene 1.0 ST Array 10895613 - -
          Affymetrix Rat Gene 1.0 ST Array 10895617 - -
          Affymetrix Rat Gene 1.0 ST Array 10895610 - -
          Affymetrix Rat Gene 1.0 ST Array 10895607 - -
          Affymetrix Rat Gene 1.0 ST Array 10895611 - -
          Affymetrix Rat Gene 1.0 ST Array 10895604 - -
          Affymetrix Rat Gene 1.0 ST Array 10895616 - -
          Affymetrix Rat Gene 1.0 ST Array 10895601 - -
          Affymetrix Rat Gene 1.1 ST Array 10895600 - -
          Rat Genome 230 2.0 Array 1389485_at - -
          Rat Genome 230 2.0 Array 1389485_PM_at - -
          Affymetrix HT Rat Focus Array 1389485_at - -
          Affymetrix HT Rat Focus Array 1389485_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700005952.rn.1 TC0700000771.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700005949.rn.1 TC0700000771.rn.1 RTA TC
          Rat Expression Set 230 1389485_at - -
          Rat Expression Set 230 1389485_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10895600 - -
          Rat Genome U34 Set rc_AI178622_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21526492 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360593 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline