Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01504446_m1
          See other TAF2 GE Assays ›
          Gene Symbol:
          Taf2
          Gene Name:
          TAF2 RNA polymerase II, TATA box binding protein (TBP)-associated factor
          Gene Aliases:
          TAFIIB
          Chromosome Location:
          Chr.7: 94698987 - 94755924 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01504446_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Taf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133319.1 NP_579853.1 25 - 26 3549 80
          XM_006241644.1 XP_006241706.1 21 - 22 2863 80
          GenBank mRNA BC091307.1 - 7 - 8 874 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003142.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          transcription initiation from RNA polymerase II promoter
          proteolysis
          response to organic cyclic compound
          positive regulation of transcription from RNA polymerase II promoter
          molecular_function
          metallopeptidase activity
          zinc ion binding
          transcription regulatory region DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P031800 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P484152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453613 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111622 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47928 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165719 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133035 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168880 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46305 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P484152 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031800 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031800 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P484152 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P484152 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P031800 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P122201 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P468312 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62807 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49257 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10903781 - -
          Affymetrix Rat Gene 1.0 ST Array 10903786 - -
          Affymetrix Rat Gene 1.0 ST Array 10903773 - -
          Affymetrix Rat Gene 1.0 ST Array 10903774 - -
          Affymetrix Rat Gene 1.0 ST Array 10903784 - -
          Affymetrix Rat Gene 1.0 ST Array 10903780 - -
          Affymetrix Rat Gene 1.0 ST Array 10903778 - -
          Affymetrix Rat Gene 1.0 ST Array 10903779 - -
          Affymetrix Rat Gene 1.0 ST Array 10903789 - -
          Affymetrix Rat Gene 1.0 ST Array 10903782 - -
          Affymetrix Rat Gene 1.0 ST Array 10903787 - -
          Affymetrix Rat Gene 1.0 ST Array 10903771 - -
          Affymetrix Rat Gene 1.0 ST Array 10903770 - -
          Affymetrix Rat Gene 1.0 ST Array 10903769 - -
          Affymetrix Rat Gene 1.0 ST Array 10903785 - -
          Affymetrix Rat Gene 1.0 ST Array 10903766 - -
          Affymetrix Rat Gene 1.0 ST Array 10903788 - -
          Affymetrix Rat Gene 1.0 ST Array 10903775 - -
          Affymetrix Rat Gene 1.0 ST Array 10903768 - -
          Affymetrix Rat Gene 1.0 ST Array 10903765 - -
          Affymetrix Rat Gene 1.0 ST Array 10903776 - -
          Affymetrix Rat Gene 1.0 ST Array 10903764 - -
          Affymetrix Rat Gene 1.0 ST Array 10903772 - -
          Affymetrix Rat Gene 1.0 ST Array 10903767 - -
          Affymetrix Rat Gene 1.0 ST Array 10903777 - -
          Affymetrix Rat Gene 1.0 ST Array 10903783 - -
          Affymetrix Rat Gene 1.1 ST Array 10903763 - -
          Rat Genome 230 2.0 Array 1392511_PM_at - -
          Rat Genome 230 2.0 Array 1392511_at - -
          Affymetrix HT Rat Focus Array 1392511_at - -
          Affymetrix HT Rat Focus Array 1392511_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700024046.rn.1 TC0700003142.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700024047.rn.1 TC0700003142.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700024048.rn.1 TC0700003142.rn.1 RTA TC
          Rat Expression Set 230 1392511_at - -
          Rat Expression Set 230 1392511_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10903763 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21537157 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368923 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline