Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01504576_m1
          See other SCX GE Assays ›
          Gene Symbol:
          Scx
          Gene Name:
          scleraxis
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 117519075 - 117521093 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01504576_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scx
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130508.1 NP_001123980.1 1 - 2 761 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001357.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mesoderm formation
          tissue homeostasis
          endochondral ossification
          chondrocyte differentiation
          heart valve morphogenesis
          heart valve formation
          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          positive regulation of cell proliferation
          positive regulation of gene expression
          cell differentiation
          collagen fibril organization
          BMP signaling pathway
          positive regulation of collagen biosynthetic process
          skeletal muscle cell differentiation
          tendon development
          tendon cell differentiation
          tendon formation
          deltoid tuberosity development
          negative regulation of apoptotic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          embryonic skeletal system development
          face morphogenesis
          regulation of cartilage development
          positive regulation of cartilage development
          sclerotome development
          cellular response to mechanical stimulus
          cellular response to transforming growth factor beta stimulus
          cellular response to BMP stimulus
          positive regulation of gastrulation
          RNA polymerase II distal enhancer sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
          DNA binding
          bHLH transcription factor binding
          sequence-specific DNA binding
          protein heterodimerization activity
          E-box binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P039737 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P142942 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142942 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039737 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P039737 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P142942 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P039737 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P142942 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10897287 - -
          Affymetrix Rat Gene 1.0 ST Array 10897286 - -
          Affymetrix Rat Gene 1.1 ST Array 10897285 - -
          Rat Genome 230 2.0 Array 1393338_at - -
          Rat Genome 230 2.0 Array 1393338_PM_at - -
          Affymetrix HT Rat Focus Array 1393338_PM_at - -
          Affymetrix HT Rat Focus Array 1393338_at - -
          Affymetrix Rat Clariom S Assay PSR0700009934.rn.1 TC0700001357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700009935.rn.1 TC0700001357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700009937.rn.1 TC0700001357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700009938.rn.1 TC0700001357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700009931.rn.1 TC0700001357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700009932.rn.1 TC0700001357.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700009933.rn.1 TC0700001357.rn.1 RTA TC
          Rat Expression Set 230 1393338_at - -
          Rat Expression Set 230 1393338_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10897285 - -
          Rat Genome U34 Set rc_AI172335_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21302340 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline