Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01505070_m1
          See other TCF20 GE Assays ›
          Gene Symbol:
          Tcf20
          Gene Name:
          transcription factor 20
          Gene Aliases:
          AR1, SPBP
          Chromosome Location:
          Chr.7: 123670135 - 123767797 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01505070_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tcf20
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130574.1 NP_001124046.1 2 - 3 5937 67
          XM_006242110.2 XP_006242172.1 1 - 2 6837 67
          XM_006242112.2 XP_006242174.1 1 - 2 6837 67
          XM_008765772.1 XP_008763994.1 1 - 2 6836 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003580.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of transcription from RNA polymerase II promoter
          transcription factor activity, sequence-specific DNA binding
          zinc ion binding
          transcription regulatory region DNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P544019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315126 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 900742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 480205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735791 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35165 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24420 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69224 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61883 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122548 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92339 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79926 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174697 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67500 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125172 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146749 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120926 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P544019 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P544019 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P544019 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P544019 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10905767 - -
          Affymetrix Rat Gene 1.0 ST Array 10905768 - -
          Affymetrix Rat Gene 1.0 ST Array 10905769 - -
          Affymetrix Rat Gene 1.1 ST Array 10905765 - -
          Rat Genome 230 2.0 Array 1393774_at - -
          Rat Genome 230 2.0 Array 1390817_at - -
          Rat Genome 230 2.0 Array 1390390_PM_at - -
          Rat Genome 230 2.0 Array 1393774_PM_at - -
          Rat Genome 230 2.0 Array 1390817_PM_at - -
          Rat Genome 230 2.0 Array 1390390_at - -
          Affymetrix HT Rat Focus Array 1390390_PM_at - -
          Affymetrix HT Rat Focus Array 1390390_at - -
          Affymetrix HT Rat Focus Array 1393774_PM_at - -
          Affymetrix HT Rat Focus Array 1393774_at - -
          Affymetrix Rat Clariom S Assay PSR0700028298.rn.1 TC0700003580.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700028299.rn.1 TC0700003580.rn.1 RTA TC
          Rat Expression Set 230 1390390_PM_at - -
          Rat Expression Set 230 1390390_at - -
          Rat Expression Set 230 1393774_PM_at - -
          Rat Expression Set 230 1393774_at - -
          Affymetrix Rat Gene 1.0 ST Array 10905765 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20724139 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368007 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline