Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01505955_m1
          See other NAB2 GE Assays ›
          Gene Symbol:
          Nab2
          Gene Name:
          Ngfi-A binding protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 70963580 - 70971480 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01505955_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nab2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134874.1 NP_001128346.1 1 - 2 294 72
          XM_006241459.2 XP_006241521.1 1 - 2 1802 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002927.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          endochondral ossification
          regulation of transcription, DNA-templated
          Schwann cell differentiation
          negative regulation of transcription from RNA polymerase III promoter
          myelination
          regulation of epidermis development
          transcription factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P144913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599876 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144913 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P144913 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P144913 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1030919 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10903175 - -
          Affymetrix Rat Gene 1.0 ST Array 10903169 - -
          Affymetrix Rat Gene 1.0 ST Array 10903168 - -
          Affymetrix Rat Gene 1.0 ST Array 10903174 - -
          Affymetrix Rat Gene 1.0 ST Array 10903173 - -
          Affymetrix Rat Gene 1.0 ST Array 10903170 - -
          Affymetrix Rat Gene 1.0 ST Array 10903171 - -
          Affymetrix Rat Gene 1.0 ST Array 10903167 - -
          Affymetrix Rat Gene 1.0 ST Array 10903172 - -
          Affymetrix Rat Gene 1.1 ST Array 10895932 - -
          Affymetrix Rat Gene 1.1 ST Array 10903166 - -
          Rat Genome 230 2.0 Array 1374925_PM_at - -
          Rat Genome 230 2.0 Array 1374925_at - -
          Rat Genome 230 2.0 Array 1391527_at - -
          Rat Genome 230 2.0 Array 1391527_PM_at - -
          Affymetrix HT Rat Focus Array 1374925_PM_at - -
          Affymetrix HT Rat Focus Array 1374925_at - -
          Affymetrix HT Rat Focus Array 1391527_PM_at - -
          Affymetrix HT Rat Focus Array 1391527_at - -
          Affymetrix Rat Clariom S Assay PSR0700022767.rn.1 TC0700002927.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700022766.rn.1 TC0700002927.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700022769.rn.1 TC0700002927.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700022770.rn.1 TC0700002927.rn.1 RTA TC
          Rat Expression Set 230 1374925_at - -
          Rat Expression Set 230 1374925_PM_at - -
          Rat Expression Set 230 1391527_PM_at - -
          Rat Expression Set 230 1391527_at - -
          Affymetrix Rat Gene 1.0 ST Array 10903166 - -
          Rat Genome U34 Set rc_AI102530_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20944674 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368836 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline