Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01507961_m1
          See other GPRC5B GE Assays ›
          Gene Symbol:
          Gprc5b
          Gene Name:
          G protein-coupled receptor, class C, group 5, member B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 188688743 - 188713709 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01507961_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gprc5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106304.1 NP_001099774.1 2 - 3 1174 90
          XM_006230109.2 XP_006230171.1 2 - 3 1148 90
          XM_006230110.2 XP_006230172.1 2 - 3 1206 90
          XM_008759715.1 XP_008757937.1 2 - 3 1221 90
          GenBank mRNA BC166986.1 - 2 - 3 1138 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006502.rn.1

          Set Membership:

          Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          G-protein coupled receptor signaling pathway
          locomotory behavior
          positive regulation of neuron projection development
          glucose homeostasis
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of neuron differentiation
          positive regulation of protein kinase activity
          positive regulation of inflammatory response
          positive regulation of macrophage cytokine production
          positive regulation of protein tyrosine kinase activity
          positive regulation of canonical Wnt signaling pathway
          G-protein coupled receptor activity
          protein kinase binding
          protein kinase activator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P535835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301269 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90766 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163043 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47065 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123888 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P535835 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P535835 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P535835 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P535835 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P773535 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P342273 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104601 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10725281 - -
          Affymetrix Rat Gene 1.0 ST Array 10725283 - -
          Affymetrix Rat Gene 1.0 ST Array 10725282 - -
          Affymetrix Rat Gene 1.1 ST Array 10725279 - -
          Rat Genome 230 2.0 Array 1373336_at - -
          Rat Genome 230 2.0 Array 1373336_PM_at - -
          Rat Genome 230 2.0 Array 1371671_at - -
          Rat Genome 230 2.0 Array 1371671_PM_at - -
          Affymetrix HT Rat Focus Array 1371671_PM_at - -
          Affymetrix HT Rat Focus Array 1373336_PM_at - -
          Affymetrix HT Rat Focus Array 1373336_at - -
          Affymetrix HT Rat Focus Array 1371671_at - -
          Affymetrix Rat Clariom S Assay PSR0100051051.rn.1 TC0100006502.rn.1 RTA TC
          Rat Expression Set 230 1373336_PM_at - -
          Rat Expression Set 230 1373336_at - -
          Rat Expression Set 230 1371671_at - -
          Rat Expression Set 230 1371671_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10725279 - -
          Rat Genome U34 Set rc_AA924526_at - -
          Rat Genome U34 Set rc_AI102424_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21700624 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372612 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline