Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01507970_g1
          See other LAMTOR1 GE Assays ›
          Gene Symbol:
          Lamtor1
          Gene Name:
          late endosomal/lysosomal adaptor, MAPK and MTOR activator 1
          Gene Aliases:
          p18
          Chromosome Location:
          Chr.1: 166991474 - 166997045 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01507970_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lamtor1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199102.1 NP_954533.1 4 - 5 455 67
          GenBank mRNA BC061783.1 - 4 - 5 455 5623933 67
          FQ214531.1 - 4 - 5 459 67
          FQ219988.1 - 4 - 5 468 67
          FQ220581.1 - 4 - 5 451 67
          FQ220693.1 - 4 - 5 453 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lamtor1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of receptor recycling
          endosome organization
          lysosome organization
          regulation of cholesterol esterification
          regulation of cholesterol efflux
          cell growth
          positive regulation of TOR signaling
          lysosome localization
          endosome localization
          cellular protein localization
          cholesterol homeostasis
          positive regulation of MAPK cascade
          positive regulation of GTPase activity
          regulation of cholesterol import
          cellular response to amino acid stimulus
          guanyl-nucleotide exchange factor activity
          protein binding
          protein complex scaffold

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P005008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832156 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131655 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P005008 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P005008 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P410272 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46220 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10709272 - -
          Affymetrix Rat Gene 1.0 ST Array 10709270 - -
          Affymetrix Rat Gene 1.0 ST Array 10709273 - -
          Affymetrix Rat Gene 1.0 ST Array 10933588 - -
          Affymetrix Rat Gene 1.0 ST Array 10709269 - -
          Affymetrix Rat Gene 1.0 ST Array 10709271 - -
          Affymetrix Rat Gene 1.1 ST Array 10709268 - -
          Affymetrix Rat Gene 1.1 ST Array 10933587 - -
          Rat Genome 230 2.0 Array 1388980_PM_at - -
          Rat Genome 230 2.0 Array 1388980_at - -
          Affymetrix HT Rat Focus Array 1388980_PM_at - -
          Affymetrix HT Rat Focus Array 1388980_at - -
          Affymetrix Rat Clariom S Assay PSR0100065459.rn.1 TC0100008050.rn.1 RTA TC
          Rat Expression Set 230 1388980_PM_at - -
          Rat Expression Set 230 1388980_at - -
          Affymetrix Rat Gene 1.0 ST Array 10709268 - -
          Affymetrix Rat Gene 1.0 ST Array 10933587 - -
          Rat Genome U34 Set rc_AA799718_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20899040 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352579 - -
          RatRef-12 Expression BeadChip ILMN_58007 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline