Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01508861_m1
          See other GGA2 GE Assays ›
          Gene Symbol:
          Gga2
          Gene Name:
          golgi associated, gamma adaptin ear containing, ARF binding protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 191951578 - 191984858 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01508861_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gga2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100519.1 NP_001093989.1 16 - 17 1778 77
          GenBank mRNA BC098781.1 - 10 - 11 1056 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006556.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          vesicle-mediated transport
          ADP-ribosylation factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P18343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195318 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173106 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37922 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59990 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18343 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18343 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P18343 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18343 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103295 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10725460 - -
          Affymetrix Rat Gene 1.0 ST Array 10725446 - -
          Affymetrix Rat Gene 1.0 ST Array 10725450 - -
          Affymetrix Rat Gene 1.0 ST Array 10725462 - -
          Affymetrix Rat Gene 1.0 ST Array 10725459 - -
          Affymetrix Rat Gene 1.0 ST Array 10725455 - -
          Affymetrix Rat Gene 1.0 ST Array 10725452 - -
          Affymetrix Rat Gene 1.0 ST Array 10725458 - -
          Affymetrix Rat Gene 1.0 ST Array 10725447 - -
          Affymetrix Rat Gene 1.0 ST Array 10725449 - -
          Affymetrix Rat Gene 1.0 ST Array 10725448 - -
          Affymetrix Rat Gene 1.0 ST Array 10725461 - -
          Affymetrix Rat Gene 1.0 ST Array 10725456 - -
          Affymetrix Rat Gene 1.0 ST Array 10725457 - -
          Affymetrix Rat Gene 1.0 ST Array 10725454 - -
          Affymetrix Rat Gene 1.0 ST Array 10725453 - -
          Affymetrix Rat Gene 1.0 ST Array 10725451 - -
          Affymetrix Rat Gene 1.1 ST Array 10725445 - -
          Rat Genome 230 2.0 Array 1372408_PM_at - -
          Rat Genome 230 2.0 Array 1395394_at - -
          Rat Genome 230 2.0 Array 1372408_at - -
          Rat Genome 230 2.0 Array 1395394_PM_at - -
          Affymetrix HT Rat Focus Array 1395394_at - -
          Affymetrix HT Rat Focus Array 1372408_PM_at - -
          Affymetrix HT Rat Focus Array 1372408_at - -
          Affymetrix HT Rat Focus Array 1395394_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100051457.rn.1 TC0100006556.rn.1 RTA TC
          Rat Expression Set 230 1372408_PM_at - -
          Rat Expression Set 230 1372408_at - -
          Rat Expression Set 230 1395394_PM_at - -
          Rat Expression Set 230 1395394_at - -
          Affymetrix Rat Gene 1.0 ST Array 10725445 - -
          Rat Genome U34 Set rc_AI030192_at - -
          Rat Genome U34 Set rc_AI177594_at - -
          Rat Genome U34 Set rc_AI176388_s_at - -
          Rat Genome U34 Set rc_AI233767_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21163553 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365996 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline