Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01510147_g1
          See other FOLR2 GE Assays ›
          Gene Symbol:
          Folr2
          Gene Name:
          folate receptor 2 (fetal)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 166915045 - 166933377 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01510147_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Folr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106283.1 NP_001099753.1 5 - 6 743 73
          XM_006229858.2 XP_006229920.1 4 - 5 623 73
          XM_006229859.2 XP_006229921.1 5 - 6 825 73
          XM_008759703.1 XP_008757925.1 4 - 5 658 73
          GenBank mRNA FQ211323.1 - 4 - 5 590 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006166.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Transporter 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          folic acid transport
          molecular_function
          folic acid binding
          folic acid transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P788740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398009 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105029 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72736 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60074 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P788740 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P788740 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P788740 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P788740 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55906 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85778 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77003 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10724115 - -
          Affymetrix Rat Gene 1.0 ST Array 10724112 - -
          Affymetrix Rat Gene 1.0 ST Array 10724113 - -
          Affymetrix Rat Gene 1.0 ST Array 10724114 - -
          Affymetrix Rat Gene 1.1 ST Array 10724111 - -
          Rat Genome 230 2.0 Array 1390348_PM_at - -
          Rat Genome 230 2.0 Array 1390348_at - -
          Affymetrix HT Rat Focus Array 1390348_PM_at - -
          Affymetrix HT Rat Focus Array 1390348_at - -
          Affymetrix Rat Clariom S Assay PSR0100049034.rn.1 TC0100006166.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049033.rn.1 TC0100006166.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049032.rn.1 TC0100006166.rn.1 RTA TC
          Rat Expression Set 230 1390348_PM_at - -
          Rat Expression Set 230 1390348_at - -
          Affymetrix Rat Gene 1.0 ST Array 10724111 - -
          Rat Genome U34 Set rc_AA955042_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20789758 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354017 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0