Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01510333_m1
          See other PALB2 GE Assays ›
          Gene Symbol:
          Palb2
          Gene Name:
          partner and localizer of BRCA2
          Gene Aliases:
          RGD1304759
          Chromosome Location:
          Chr.1: 192064586 - 192088547 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01510333_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Palb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001191603.1 NP_001178532.1 12 - 13 3126 73
          XM_008759711.1 XP_008757933.1 10 - 11 4302 73
          XM_008759712.1 XP_008757934.1 11 - 12 3210 73
          XM_008759713.1 XP_008757935.1 11 - 12 3120 73
          GenBank mRNA FQ211178.1 - 12 - 13 3275 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006559.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via homologous recombination
          somitogenesis
          inner cell mass cell proliferation
          mesoderm development
          biological_process
          organ morphogenesis
          post-anal tail morphogenesis
          negative regulation of apoptotic process
          molecular_function
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1021489 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1021489 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1021489 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1021489 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1021489 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10725482 - -
          Affymetrix Rat Gene 1.0 ST Array 10725484 - -
          Affymetrix Rat Gene 1.0 ST Array 10725485 - -
          Affymetrix Rat Gene 1.0 ST Array 10725486 - -
          Affymetrix Rat Gene 1.0 ST Array 10725492 - -
          Affymetrix Rat Gene 1.0 ST Array 10725490 - -
          Affymetrix Rat Gene 1.0 ST Array 10725493 - -
          Affymetrix Rat Gene 1.0 ST Array 10725481 - -
          Affymetrix Rat Gene 1.0 ST Array 10725487 - -
          Affymetrix Rat Gene 1.0 ST Array 10725488 - -
          Affymetrix Rat Gene 1.0 ST Array 10725483 - -
          Affymetrix Rat Gene 1.0 ST Array 10725489 - -
          Rat Genome 230 2.0 Array 1376695_at - -
          Rat Genome 230 2.0 Array 1376695_PM_at - -
          Affymetrix HT Rat Focus Array 1376695_PM_at - -
          Affymetrix HT Rat Focus Array 1376695_at - -
          Affymetrix Rat Clariom S Assay PSR0100051511.rn.1 TC0100006559.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100051512.rn.1 TC0100006559.rn.1 RTA TC
          Rat Expression Set 230 1376695_PM_at - -
          Rat Expression Set 230 1376695_at - -
          Affymetrix Rat Gene 1.0 ST Array 10725480 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21954213 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368829 - -
          RatRef-12 Expression BeadChip ILMN_53548 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline