Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01510663_g1
          See other ART5 GE Assays ›
          Gene Symbol:
          Art5
          Gene Name:
          ADP-ribosyltransferase 5
          Gene Aliases:
          ARTC5
          Chromosome Location:
          Chr.1: 167192805 - 167202767 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01510663_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Art5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013039.1 NP_001013057.1 5 - 6 1619 61
          XM_006229843.2 XP_006229905.1 3 - 4 1205 61
          XM_006229845.2 XP_006229907.1 3 - 4 1207 61
          XM_006229846.2 XP_006229908.1 4 - 5 1237 61
          XM_008759692.1 XP_008757914.1 3 - 4 1149 61
          GenBank mRNA BC083915.1 - 5 - 6 1619 7134556 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100006176.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Art5

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ADP-ribosylation
          NAD+ ADP-ribosyltransferase activity
          NAD+ nucleosidase activity
          NAD(P)+-protein-arginine ADP-ribosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P086096 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P042586 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105215 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71126 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042586 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P086096 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042586 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P086096 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P042586 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P086096 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P407362 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23945 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91070 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10724155 - -
          Affymetrix Rat Gene 1.0 ST Array 10724159 - -
          Affymetrix Rat Gene 1.0 ST Array 10724156 - -
          Affymetrix Rat Gene 1.0 ST Array 10724163 - -
          Affymetrix Rat Gene 1.0 ST Array 10724162 - -
          Affymetrix Rat Gene 1.0 ST Array 10724158 - -
          Affymetrix Rat Gene 1.0 ST Array 10724157 - -
          Affymetrix Rat Gene 1.0 ST Array 10724161 - -
          Affymetrix Rat Gene 1.0 ST Array 10724154 - -
          Affymetrix Rat Gene 1.0 ST Array 10724160 - -
          Affymetrix Rat Gene 1.1 ST Array 10724153 - -
          Rat Genome 230 2.0 Array 1391268_PM_at - -
          Rat Genome 230 2.0 Array 1391268_at - -
          Affymetrix HT Rat Focus Array 1391268_at - -
          Affymetrix HT Rat Focus Array 1391268_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100049134.rn.1 TC0100006176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049133.rn.1 TC0100006176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049137.rn.1 TC0100006176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049138.rn.1 TC0100006176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049136.rn.1 TC0100006176.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100049139.rn.1 TC0100006176.rn.1 RTA TC
          Rat Expression Set 230 1391268_PM_at - -
          Rat Expression Set 230 1391268_at - -
          Affymetrix Rat Gene 1.0 ST Array 10724153 - -
          Rat Genome U34 Set rc_AI070764_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22348524 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362379 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline