Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01516455_m1
          See other SF3A3 GE Assays ›
          Gene Symbol:
          Sf3a3
          Gene Name:
          splicing factor 3a, subunit 3
          Gene Aliases:
          LRRGT00093
          Chromosome Location:
          Chr.5: 142702721 - 142722350 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01516455_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sf3a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025698.1 NP_001020869.1 12 - 13 1067 103
          GenBank mRNA BC099183.1 - 12 - 13 1067 7383264 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001430.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sf3a3

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          mRNA processing
          RNA binding
          zinc ion binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P18203 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P131726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182227 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114750 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173871 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18203 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P131726 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P131726 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P18203 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P18203 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P131726 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P18203 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100352 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40057 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10871824 - -
          Affymetrix Rat Gene 1.0 ST Array 10871814 - -
          Affymetrix Rat Gene 1.0 ST Array 10871828 - -
          Affymetrix Rat Gene 1.0 ST Array 10871823 - -
          Affymetrix Rat Gene 1.0 ST Array 10871819 - -
          Affymetrix Rat Gene 1.0 ST Array 10871816 - -
          Affymetrix Rat Gene 1.0 ST Array 10871830 - -
          Affymetrix Rat Gene 1.0 ST Array 10871826 - -
          Affymetrix Rat Gene 1.0 ST Array 10871815 - -
          Affymetrix Rat Gene 1.0 ST Array 10871817 - -
          Affymetrix Rat Gene 1.0 ST Array 10871825 - -
          Affymetrix Rat Gene 1.0 ST Array 10871818 - -
          Affymetrix Rat Gene 1.0 ST Array 10871822 - -
          Affymetrix Rat Gene 1.0 ST Array 10871829 - -
          Affymetrix Rat Gene 1.0 ST Array 10871820 - -
          Affymetrix Rat Gene 1.0 ST Array 10871827 - -
          Affymetrix Rat Gene 1.1 ST Array 10871813 - -
          Rat Genome 230 2.0 Array 1388669_at - -
          Rat Genome 230 2.0 Array 1388669_PM_at - -
          Affymetrix HT Rat Focus Array 1388669_at - -
          Affymetrix HT Rat Focus Array 1388669_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500009928.rn.1 TC0500001430.rn.1 RTA TC
          Rat Expression Set 230 1388669_PM_at - -
          Rat Expression Set 230 1388669_at - -
          Affymetrix Rat Gene 1.0 ST Array 10871813 - -
          Rat Genome U34 Set rc_AA818612_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356020 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline