Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01520131_m1
          See other MTFR1L GE Assays ›
          Gene Symbol:
          Mtfr1l
          Gene Name:
          mitochondrial fission regulator 1-like
          Gene Aliases:
          Fam54b, RGD1305453
          Chromosome Location:
          Chr.5: 152736580 - 152746686 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01520131_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mtfr1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013935.1 NP_001013957.1 6 - 7 893 87
          XM_006239124.2 XP_006239186.1 6 - 7 1189 87
          XM_006239125.2 XP_006239187.1 6 - 7 967 87
          GenBank mRNA BC083692.1 - 6 - 7 893 7189448 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003816.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mtfr1l

          Gene Ontology Categories:

          Function(s)

          mitochondrial fission
          aerobic respiration

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P107549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417081 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111818 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P107549 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P107549 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P107549 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95537 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16975 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10880520 - -
          Affymetrix Rat Gene 1.0 ST Array 10880517 - -
          Affymetrix Rat Gene 1.0 ST Array 10880521 - -
          Affymetrix Rat Gene 1.0 ST Array 10880518 - -
          Affymetrix Rat Gene 1.0 ST Array 10880523 - -
          Affymetrix Rat Gene 1.0 ST Array 10880522 - -
          Affymetrix Rat Gene 1.0 ST Array 10880519 - -
          Affymetrix Rat Gene 1.1 ST Array 10880516 - -
          Rat Genome 230 2.0 Array 1371461_at - -
          Rat Genome 230 2.0 Array 1371461_PM_at - -
          Rat Genome 230 2.0 Array 1385293_PM_at - -
          Rat Genome 230 2.0 Array 1385293_at - -
          Affymetrix HT Rat Focus Array 1385293_at - -
          Affymetrix HT Rat Focus Array 1371461_PM_at - -
          Affymetrix HT Rat Focus Array 1385293_PM_at - -
          Affymetrix HT Rat Focus Array 1371461_at - -
          Affymetrix Rat Clariom S Assay PSR0500028619.rn.1 TC0500003816.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028621.rn.1 TC0500003816.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500028620.rn.1 TC0500003816.rn.1 RTA TC
          Rat Expression Set 230 1371461_at - -
          Rat Expression Set 230 1371461_PM_at - -
          Rat Expression Set 230 1385293_at - -
          Rat Expression Set 230 1385293_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10880516 - -
          Rat Genome U34 Set rc_AI007826_at - -
          Rat Genome U34 Set rc_AI045822_s_at - -
          Rat Genome U34 Set rc_AI172255_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21696950 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354229 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline