Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01520674_m1
          See other RPA2 GE Assays ›
          Gene Symbol:
          Rpa2
          Gene Name:
          replication protein A2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 150957041 - 150967597 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01520674_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpa2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021582.1 NP_067593.1 8 - 9 811 63
          GenBank mRNA BC079180.1 - 8 - 9 811 7130711 63
          FQ228766.1 - 5 - 6 440 63
          X98490.1 - 7 - 8 717 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001609.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpa2

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          double-strand break repair via homologous recombination
          DNA replication
          base-excision repair
          nucleotide-excision repair
          mismatch repair
          regulation of double-strand break repair via homologous recombination
          mitotic G1 DNA damage checkpoint
          regulation of DNA damage checkpoint
          DNA binding
          damaged DNA binding
          single-stranded DNA binding
          enzyme binding
          protein phosphatase binding
          ubiquitin protein ligase binding
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P641107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 833128 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109050 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 131581 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P641107 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P641107 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P641107 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P641107 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700373695 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51427 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94105 - -
          Agilent Rat cDNA Microarray (G4105A) 700373695 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10880367 - -
          Affymetrix Rat Gene 1.0 ST Array 10880365 - -
          Affymetrix Rat Gene 1.0 ST Array 10880362 - -
          Affymetrix Rat Gene 1.0 ST Array 10880366 - -
          Affymetrix Rat Gene 1.0 ST Array 10880363 - -
          Affymetrix Rat Gene 1.0 ST Array 10880370 - -
          Affymetrix Rat Gene 1.0 ST Array 10880369 - -
          Affymetrix Rat Gene 1.0 ST Array 10880371 - -
          Affymetrix Rat Gene 1.0 ST Array 10880364 - -
          Affymetrix Rat Gene 1.0 ST Array 10880368 - -
          Affymetrix Rat Gene 1.1 ST Array 10880361 - -
          Rat Genome 230 2.0 Array 1388135_at - -
          Rat Genome 230 2.0 Array 1388135_PM_at - -
          Affymetrix HT Rat Focus Array 1388135_at - -
          Affymetrix HT Rat Focus Array 1388135_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500011567.rn.1 TC0500001609.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011569.rn.1 TC0500001609.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011568.rn.1 TC0500001609.rn.1 RTA TC
          Rat Expression Set 230 1388135_PM_at - -
          Rat Expression Set 230 1388135_at - -
          Affymetrix Rat Gene 1.0 ST Array 10880361 - -
          Rat Genome U34 Set X98490cds_at - -
          Rat Genome U34 Set X98490cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21762788 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359929 - -
          RatRef-12 Expression BeadChip ILMN_68423 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline